Skip to Content
Merck
All Photos(1)

Key Documents

H2529

Sigma-Aldrich

α-Hydroxytriazolam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12Cl2N4O
CAS Number:
Molecular Weight:
359.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

Quality Level

SMILES string

OCc1nnc2CN=C(c3ccccc3Cl)c4cc(Cl)ccc4-n12

InChI

1S/C17H12Cl2N4O/c18-10-5-6-14-12(7-10)17(11-3-1-2-4-13(11)19)20-8-15-21-22-16(9-24)23(14)15/h1-7,24H,8-9H2

InChI key

BHUYWUDMVCLHND-UHFFFAOYSA-N

Biochem/physiol Actions

Triazolam metabolite

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin, L., et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 654, 77-77 (1994)
A D Fraser et al.
Journal of analytical toxicology, 16(6), 347-350 (1992-11-01)
Triazolam is a very short-acting triazolobenzodiazepine with sedative-hypnotic properties. Approximately 2% of an oral dose is excreted unchanged in the urine. The major urinary metabolite is alpha-hydroxytriazolam glucuronide (70% of the dose). The objective of this study was to characterize
Fumio Moriya et al.
Legal medicine (Tokyo, Japan), 5 Suppl 1, S91-S95 (2003-08-26)
A 57-year-old man was found dead lying down in a bamboo thicket. Moderate to severe petechiae were present on his conjunctivae, buccal mucosa, and laryngeal mucosa at autopsy. Cardiac chambers contained a normal volume of fluid blood. Moderate atherosclerosis and
Dora Farkas et al.
Journal of clinical pharmacology, 47(3), 286-294 (2007-02-27)
The effect of pomegranate juice (PJ) or grapefruit juice (GFJ) on CYP3A activity was studied in vitro and in healthy human volunteers. In human liver microsomes, the mean 50% inhibitory concentrations (IC(50)) for PJ and GFJ versus CYP3A (triazolam alpha-hydroxylation)
Qiran Sun et al.
Journal of analytical toxicology, 34(2), 89-94 (2010-03-13)
A sensitive liquid chromatography-tandem mass spectrometry method is presented for determination of triazolam and alpha-hydroxytriazolam in guinea pig hair after a single dose of triazolam. Eighteen guinea pigs were divided into three dosage groups (10, 100, and 500 microg/kg) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service