Skip to Content
Merck
All Photos(1)

Key Documents

GW22560F

Sigma-Aldrich

Anti-SH3GL1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SH3GL1(6455)

General description

SH3GL1 (SH3-domain GRB2-like 1), also called EA2 (endophilin A2), was initially recognized as a fusion partner of MLL (mixed-lineage leukaemia) translocation in childhood leukaemia. It is the first member of the EEN/EA SH3 domain (Src homology 3 domain)-containing family of proteins. It is found in the cells of haemopoietic, fibroblast and epithelial lineage, where it resides primarily in the nucleus. In neurons and osteoclasts, its primary localization is in cytoplasm, thus suggesting its shuttles between nucleus and cytoplasm.

Immunogen

Immunogen Sequence: GI # 4506929, sequence 1-360
Recombinant SH3-domain GRB2-like 1

Application

Anti-SH3GL1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

SH3GL1 (SH3-domain GRB2-like 1) or endophilin-A2 is involved in the formation of endocytic vesicles. The protein is a 50- to 60-amino-acid module carried by intracellular proteins involved in the transduction of signals for cell polarization, motility, enzymatic activation, and transcriptional regulation. SH3GL1 is expressed ubiquitously. It is one of the main accessory proteins involved in clathrin-mediated endocytosis and it interacts with other endocytic proteins, such as dynamin, by its SH3 domain. SH3GL1 may be one of the pathogenic genes for idiopathic scoliosis (IS) and may be associated with adolescent idiopathic scoliosis (AIS). It may have some role in the cell cycle. SH3GL1 may be involved in the oncogenic process of gliomas.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jifeng Zhang et al.
Cellular signalling, 24(11), 2043-2050 (2012-07-04)
Endophilin, one of the main accessory proteins involved in clathrin-mediated endocytosis, interacts with other endocytic proteins, such as dynamin, by its SH3 domain. We previously reported that voltage-gated Ca(2+) channels are an integral part of the synaptic vesicle (SV) endocytosis
Ngai Cheung et al.
The Biochemical journal, 383(Pt 1), 27-35 (2004-06-25)
EEN (extra eleven nineteen), also known as EA2 (endophilin A2), a fusion partner of the MLL (mixed-lineage leukaemia) gene in human acute leukaemia, is a member of the endophilin A family, involved in the formation of endocytic vesicles. We present
Tao Yang et al.
Zhonghua wai ke za zhi [Chinese journal of surgery], 48(6), 435-438 (2010-07-16)
To identify whether SH3GL1 gene serves as a disease associated gene of adolescent idiopathic scoliosis (AIS). Positioning candidate cloning: "case-sibling or case-family control design" research scheme based on family constellation was designed. Fifty-six AIS patients (15 male and 41 female
Tomoo Matsutani et al.
Journal of experimental & clinical cancer research : CR, 31, 85-85 (2012-10-12)
Glioma is the most common primary malignant central nervous system tumor in adult, and is usually not curable in spite of various therapeutic approaches. Clarification of the oncogenic process in its early stage is important for the diagnosis and effective
Tao Yang et al.
International orthopaedics, 36(6), 1307-1314 (2011-12-21)
To investigate the effects of genetic factors on idiopathic scoliosis (IS) and genetic modes through genetic epidemiological survey on IS in Chongqing City, China, and to determine whether SH3GL1, GADD45B, and FGF22 in the chromosome 19p13.3 are the pathogenic genes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service