Skip to Content
Merck
All Photos(1)

Key Documents

GW21229

Sigma-Aldrich

Anti-CLIF antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Cycle-like factor CLIF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21229

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLIF(56938)

Immunogen

Immunogen Sequence: GI # 9910368, sequence 8-100
Recombinant transcription factor BMAL2

Application

Anti-CLIF antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

The CLIF gene encodes a member of the basic helix-loop-helix PER-ARNT-SIM family of transcription factors and is located on chromosome12. It helps in controlling diverse physiological processes including circadian rhythm. CLIF is expressed in endothelial cells and neurons in the brain, including the suprachiasmatic nucleus and the center of the circadian clock. In endothelial cells, it forms a heterodimer with CLOCK and up-regulates the PAI-1 gene through E-box sites. CLIF helps in regulating the circadian oscillation of PAI-1 gene expression in endothelial cells. Its expression is increased in colorectal cancer and is related to tumor invasiveness and aggressiveness.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ikeda et al.
Biochemical and biophysical research communications, 275(2), 493-502 (2000-08-31)
We isolated a human cDNA encoding a novel member of the bHLH-PAS transcription factor superfamily, BMAL2, which is highly similar to, but distinct from, BMAL1. The composite cDNA covered a 1720-bp sequence consisting of a putative 1653-bp open reading frame
John A Schoenhard et al.
American journal of physiology. Cell physiology, 283(1), C103-C114 (2002-06-11)
The BMAL2 gene encodes a member of the basic helix-loop-helix PER-ARNT-SIM family of transcription factors, which control diverse physiological processes including circadian rhythms. We identified four novel human BMAL2 transcripts that differ by alternative splicing within their NH2-terminal regions. Divergent
Gianluigi Mazzoccoli et al.
Journal of cancer research and clinical oncology, 138(3), 501-511 (2011-12-27)
Cathepsin and plasmin may favor cancer cell invasion degrading extracellular matrix. Plasmin formation from plasminogen is regulated by plasminogen activator inhibitor type-1 (PAI-1). ARNTL2 activates the promoters of the PAI-1 gene, officially called SERPINE1, driving the circadian variation in circulating
K Maemura et al.
The Journal of biological chemistry, 275(47), 36847-36851 (2000-10-06)
The onset of myocardial infarction occurs frequently in the early morning, and it may partly result from circadian variation of fibrinolytic activity. Plasminogen activator inhibitor-1 activity shows a circadian oscillation and may account for the morning onset of myocardial infarction.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service