GEUS78220
illustra™ ExoProStar™
Cytiva US78220, sufficient for 20 reactions
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106300
NACRES:
NA.52
Recommended Products
usage
sufficient for 20 reactions
manufacturer/tradename
Cytiva US78220
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Enzymatic PCR clean up technology with illustra™ Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes.illustra™ ExoProStar is a new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits! The ExoProStar PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with illustra™ Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra™ ExoProStar improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.
Features and Benefits
- Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
- Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
- Fast 30 min protocol
- Scalable for different reaction sizes
- No loss of PCR product
- Easy to automate
- Complete heat inactivation of the enzymes within 15 min
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Legal Information
ExoProStar is a trademark of Cytiva
illustra is a trademark of Cytiva
Storage Class Code
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service