GERPN1231-2ML
Streptavidin-HRP Conjugate
Cytiva RPN1231-2ML
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.46
Recommended Products
packaging
pkg of 2 mL
manufacturer/tradename
Cytiva RPN1231-2ML
color
Colorless
storage temp.
2-8°C
Related Categories
General description
Streptavidin-Horseradish Peroxidase Conjugate.
Application
The conjugation of horseradish peroxidase and streptavidin is achieved using the periodate coupling method described by Nakane and co-workers.
This product is sold on the basis of titer in an ELISA and performance in a western blot. As a result the final Protein concentration of the conjugate (RPN 1231) will vary only minimally between batches.
This product is sold on the basis of titer in an ELISA and performance in a western blot. As a result the final Protein concentration of the conjugate (RPN 1231) will vary only minimally between batches.
Features and Benefits
- Streptavidin conjugated to reporter enzyme molecules for use in blotting and in situ hybridization applications.
- High specificity reagents.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Analysis Note
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service