GE28-9889-51
XK 26/100 Column
Synonym(s):
Affinity chromatography column, Chromatography column, Protein separation column
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115711
NACRES:
SB.52
Recommended Products
technique(s)
LPLC: suitable
Related Categories
General description
XK empty columns are designed for liquid chromatography at low to medium pressure. The user-friendly design ensures trouble-free operation with fittings for direct connection to ÄKTA design instruments or other high-performance liquid chromatography systems. The product family is based on a robust design for high reproducibility and accurate results. XK columns are compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules.
Features and Benefits
Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Column Packing and Preparation for Size Exclusion Chromatography
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service