Recommended Products
technique(s)
LPLC: suitable
Related Categories
General description
XK empty columns are designed for liquid chromatography at low to medium pressure. The user-friendly design ensures trouble-free operation with fittings for direct connection to ÄKTA design instruments or other high-performance liquid chromatography systems. The product family is based on a robust design for high reproducibility and accurate results. XK columns are compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules.
Features and Benefits
Adapter QuickLock mechanism that facilitates column handling and cleaning
Cooling jacket for sensitive purifications
Fittings for direct connection to chromatographiy systems
Plunger design to ensure a homogeneous buffer distribution and reproducible packing
User friendly and robust empty columns, designed for liquid chromatography at low to medium pressure.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Protocols
Column Packing and Preparation for Size Exclusion Chromatography