Skip to Content
Merck
All Photos(1)

Key Documents

GE28-9673-90

His Mag Sepharose Ni

Cytiva 28-9673-90, pack of 5 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56

Pricing and availability is not currently available.

ligand

IMAC immobilized with Ni

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 28-9673-90

parameter

Room temperature temp. range

matrix

paramagnetic, spherical, highly cross-linked agarose particles

capacity

50 mg binding capacity(histidine-tagged protein/ml medium)

General description

His Mag Sepharose Ni is magnetic beads designed to simplify small-scale purification and expression screening of histidine-tagged proteins.

Application

His Mag Sepharose Ni products are magnetic beads designed for rapid, small-scale purification and screening of histidine-tagged Proteins from different sources. Mag Sepharose format has excellent properties for small-scale experiments. The high density of the beads allows rapid capture by magnetic devices while the visibility of the beads ensures reliable collection of the bound histidine-tagged Proteins in the purification procedure.

The product is based on IMAC Sepharose medium with magnetite incorporated and nickel immobilized.

Together with MagRack 6, a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel.

Features and Benefits

  • High capacity magnetic beads for small-scale purification and screening of histidine-tagged Proteins
  • Easy to use visible and dense magnetic beads
  • High purity and yield
  • Easy parallel screening with high repeatability
  • Scalable-simple capture of target Protein from small μL to high mL sample volumes

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Articles

    This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

    This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

    This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

    This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

    See All

    Protocols

    This page shows magnetic bead-based purification and screening using His Mag Sepharose Ni from Cytiva.

    Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.

    Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.

    Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.

    See All

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service