GE28-9365-49
HiPrep™ Heparin Fast Flow 16/10
Cytiva 28-9365-49
Synonym(s):
Heparin affinity chromatography
About This Item
Recommended Products
ligand
porcine heparin
form
prepacked column
crosslinking
cross-linked
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 28-9365-49
storage condition
(20% ethanol containing 0.05 M sodium acetate)
parameter
1.5 bar (22 psi) (Over the Packed Bed During Operation)
2 ml/min - 10 ml/min flow rate
22 psi
technique(s)
liquid chromatography (LC): suitable
bed size
16 mm × 100 mm
bed volume
20 mL
column I.D.
16 mm
matrix
6% cross-linked agarose
matrix active group
heparin
particle size
45-165 μm
average diameter
90 μm
cleaning
4-13
working range
4-12
suitability
suitable for bioprocess medium
separation technique
affinity
Related Categories
General description
Application
Heparin is a mucopolysaccharide consisting of a repeating dimer of α-L-dipyranuronic acid 2-sulfate, and 2-deoxy-2-sulfamino-α-D-glucopyranose 6-sulfate, with a molecular weight range of 5 000 to 30 000.
Features and Benefits
- 20 mL column optimized for convenient scale-up purification .
- Immobilized heparin is used for purification of antithrombin III, coagulation factors, and other plasma Proteins.
- Simple operation with single pump configurations, as well as ÄKTA® design systems.
- Excellent flow characteristics.
Storage and Stability
Analysis Note
Other Notes
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This page shows how to remove serine proteases, e.g. thrombin and trypsin, and zymogens from recombinant products using HiTrap Benzamidine FF and Benzamidine Sepharose 4 Fast Flow from Cytiva.
Protocols
This page shows how to purify or remove DNA-binding proteins with Heparin Sepharose High Performance, Heparin Sepharose 6 Fast Flow, Capto Heparin from Cytiva.
This page shows how to use heparin in the separation of DNA binding proteins used in HiTrap Heparin HP, HiPrep 16/10 Heparin FF and Heparin Sepharose 6 Fast Flow products from Cytiva.
Affinity chromatography tags like GST fusion proteins facilitate separations in various purification products from Cytiva.
Affinity chromatography tags like GST fusion proteins facilitate separations in various purification products from Cytiva.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service