GE17-6002-45
Immobiline® Drystrip pH 3-10 NL, 24 cm
Cytiva 17-6002-45, pack of 12 strips
About This Item
Recommended Products
form
solid
feature
wetted part: no
packaging
pack of 12 strips
manufacturer/tradename
Cytiva 17-6002-45
color
Colorless
solubility
cold water: soluble (Insoluble in cold and hot water)
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Application
The pH 3–11 NL strip covers most Proteins found in prokaryotic and eukaryotic cells while the narrow-range strips can be used when higher resolution is required. The pH gradients at the extreme ends of the pH scale are nonlinear to distribute the Proteins evenly over the gel length to obtain maximal resolution.
Immobiline® DryStrip gels are rehydrated with a matching IPG Buffer prior to use. Samples can be included directly in the rehydration solution for efficient loading of dilute samples or for loading larger amounts of Protein. Samples can also be loaded after rehydration. The first-dimension separation is carried out using Ettan™ IPGphor 3 IEF system or Multiphor II flatbed system.
Features and Benefits
- Maximize resolution, loading capacity, and reproducibility of first-dimension isoelectric focusing (IEF).
- Choose from a comprehensive range of overlapping Immobiline® DryStrip gels that cover narrow (1 pH unit), medium (3–5 pH units), and broad (8 pH units) pH ranges.
- Select the broad pH 3–11 NL gradient Immobiline® DryStrip gel for fast and efficient screening to gain a broad overview of total Protein distribution.
- For more detailed studies or when using greater Protein loads, choose from four new overlapping strips covering the pH 3 to 11 range.
- Higher resolution can be obtained using narrow-range (1 pH unit) Immobiline® DryStrip gels.
- Use matching IPG buffers for optimized running conditions.
- Use IPG strips directly after reswelling without further manipulation.
- Barcoded strips for convenient sample and batch tracking.
Storage and Stability
Analysis Note
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
2-D Clean-Up Kit from Cytiva is designed to prepare samples that would otherwise produce poor 2-D results.
This page describes how to apply equilibrated Immobiline® DryStrip gels on the ExcelGel SDS gels.
This protocol shows how to prepare the Immobiline® DryStrip Kit and electrodes strips.
This page shows how to apply higher sample volumes and protein amounts with paper bridges in isoelectric focusing with Cytiva products
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service