GE17-5131-01
GSTrap™ Fast Flow
Cytiva 17-5131-01, pack of 1 × 5 mL
About This Item
Recommended Products
ligand
glutathione and 10-carbon linker arm
packaging
pack of 1 × 5 mL
manufacturer/tradename
Cytiva 17-5131-01
storage condition
(20% Ethanol)
parameter
<20 mL/min flow rate
42 psi
bed size
16 mm × 25 mm
bed volume
5 mL
column I.D.
16 mm
matrix
highly cross-linked 4% agarose
average diameter
90 μm
cleaning
3-12
working range
3-12
capacity
>10 mg/mL binding capacity
storage temp.
2-8°C
Related Categories
General description
Application
Features and Benefits
- Fast and simple one-step purification of GST-tagged Proteins.
- Mild elution conditions preserving Protein antigenicity and function.
- Easy scale-up by connecting the columns in series.
- Sepharose™ 4 Fast FLow provides good fLow properties.
Storage and Stability
Analysis Note
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This page shows troubleshooting strategies for cleavage methods using Cytiva products.
Protocols
Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.
Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.
Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.
Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service