Skip to Content
Merck
All Photos(1)

Key Documents

GE17-1287-01

Anx Seph 4 Fast Flow High Sub

Cytiva 17-1287-01, pack of 500 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

ligand

diethylaminopropyl

description

Ion Exchanger Type (value)

packaging

pack of 500 mL

manufacturer/tradename

Cytiva 17-1287-01

matrix

4% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning in place

2-14

working range

3-13

suitability

suitable for bioprocess medium

General description

ANX Sepharose 4 Fast FLow (high sub) is a weak anion exchanger, with a different selectivity compared with traditional ion exchangers. It is optimized for the separation of larger proteins.

Application

ANX Sepharose Fast FLow (high sub) is part of the Sepharose Fast FLow ion exchange platform, which has been the industrial standard for ion exchange chromatography during recent decades. ANX Sepharose 4 Fast FLow (high sub) is a weak anion exchanger medium with tertiary amine groups attached to the base matrix Sepharose 4 Fast FLow via ether linkage and a hydrophilic spacer arm. The diethylaminopropylgroup is coupled in a mode that precludes formation of quaternary groups, which are typically seen on traditional DEAE media, resulting in a truly weak anion exchanger. The base matrix, Sepharose 4 Fast Flow, is a highly cross-linked, 4% Agarose derivative with high chemical and physical stability and broad separation range. ANX Sepharose 4 Fast FLow (high sub) has high chemical stability, allowing well proven cleaning-in-place (CIP) and sanitization protocols.

As member of the BioProcess media range, ANX Sepharose 4 Fast FLow (high sub) meets industrial demands with security of supply and comprehensive technical and regulatory support.

Features and Benefits

  • Weak anion exchanger with different selectivity compared with traditional ion exchangers.
  • Applicable for separation of high molecular weight Proteins
  • Developed in co-operation with leading large-scale pharmaceutical manufacturers
  • High chemical stability allows for well proven CIP and sanitization protocols
  • The hydrophilic nature of the base matrix ensures Low levels of non-specific binding leading to Low levels of host cell-derived impurities in the elution pool.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

This page covers practical problems that may lead to a non-ideal IEX separation and their solutions.

This page covers detailed aspects of each step in an IEX separation to improve resolution and overall performance.

Protocols

This page covers the use of Sepharose Fast Flow for purification of proteins.

This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.

This page clarifies sample preparation, buffer exchange and desalting, removal of lipoproteins, phenol red, and low molecular weight contaminants in Ion exchange chromatography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service