Skip to Content
Merck
All Photos(1)

Key Documents

GE17-0300-50

Ficoll® PM 400

Cytiva 17-0300-50, pack of 500 g

Synonym(s):

Ficoll Density Gradient

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

Pricing and availability is not currently available.

form

solid
spray-dried

packaging

pack of 500 g

manufacturer/tradename

Cytiva 17-0300-50

color

White

solubility

soluble

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
F4375GE17-0300-05GE17-0300-10
form

solid, spray-dried

form

powder, spray-dried

form

solid, spray-dried

form

solid, spray-dried

color

White

color

white to off-white

color

White

color

White

packaging

pack of 500 g

packaging

-

packaging

pack of 5 kg

packaging

pack of 100 g

manufacturer/tradename

Cytiva 17-0300-50

manufacturer/tradename

-

manufacturer/tradename

Cytiva 17-0300-05

manufacturer/tradename

Cytiva 17-0300-10

General description

High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. The molecules are highly branched and the high content of hydroxyl groups leads to very good solubility in aqueous media.

Ficoll® PM400 is a synthetic neutral, highly-branched hydrophilic polymer of sucrose with an average molecular weight of 400 000. It has long been used to form density gradients for separating and isolating eukaryotic cells, organelles and bacterial cells, as a stabilizing agent, and as a preparation medium for isolating mononuclear cells. Applications can also be found in defined culture media, nucleic acid hybridization, electrophoresis, and immunological studies.
It is delivered as a spray-dried powder and is therefore readily soluble in aqueous media when it is added slowly with concomitant stirring. Any lumps formed during this procedure will dissolve fairly quickly. Concentrations up to 50% (w/v) can easily be attained

Application

Used in the formation of density gradients for the separation and isolation of eukaryotic cells, organelles, and bacterial cells, as a stabilizing agent and as a separation medium for the isolation of lymphocytes.
It also has applications in defined culture media, nucleic acid hybridization, electrophoresis, and immunological studies.
Used for gradient centrifugation in all types of centrifuge rotors and for separation at unit gravity.

Features and Benefits

  • For producing density gradients for separation of cells and subcellular components by centrifugation or by sedimentation at unit gravity.
  • Neutral, highly branched, hydrophilic polymer of sucrose which dissolves readily in aqueous solution.
  • Concentrations of up to 50% (w/v) covering densities of up to 1.2 g/mL can be achieved.
  • Better osmotic properties than sucrose.
  • Preserves functional and morphological integrity.
  • Useful for separating cells that are sensitive to centrifugation and for separating cells of similar density but different size (under conditions of sedimentation at unit gravity).
  • Does not penetrate biological membranes.
  • Serves as the raw material for preparation of Ficoll-Paque gradients.
  • Used in other applications such as electrophoresis, hybridization, cryopreservation, and as a hapten carrier.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C ()

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Ficoll is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Articles

    The table was compiled to assist the researcher in selecting references most likely to contain relevant information regarding use of Percoll in various cell types.

    Density Marker Beads facilitate Percoll gradient monitoring in biological separations.

    This page shows methods to remove Percoll from a sample after centrifugation.

    Density Marker Beads facilitate Percoll gradient monitoring in biological separations.

    See All

    Protocols

    The following procedure has been evaluated with Ficoll-Paque PLUS and is recommended for separation of normal blood samples for maximum reproducibility.

    Related Content

    Data was compiled to assist the researcher in selecting references most likely to contain relevant information regarding use of Percoll for a particular cell or tissue type.

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service