GE10600123
Amersham™ Hybond® LFP Western blotting membrane sandwich, PVDF
pore size 0.2 μm, sheet W × L 80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers, pkg of 10 ea
Synonym(s):
blotting and detection, blotting membrane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
Recommended Products
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600123
sheet W × L
80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers
pore size
0.2 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Amersham Hybond LFP PVDF membranes are optimized for fluorescent detection in Western blotting applications and for use with the Amersham ECL Plex detection system. This membrane has low background fluorescence, resulting in high sensitivity. Typical protein binding capacity is > 200 µg/cm2. Sandwich packs are designed for convenience and ease of use.
Features and Benefits
- Contains 10 Western blotting sandwiches, consisting of precut Amersham Hybond LFP 0.2 µm PVDF membranes preassembled with 2 × 3MM Chr Blotting papers.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service