GE10600015
Amersham™ Protran® Supported Western blotting membranes, nitrocellulose
pore size 0.2 μm, roll W × L 300 mm × 4 m, pkg of 1 ea
Synonym(s):
Nitrocellulose Blotting Membrane, Supported Nitrocellulose Membrane
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 10600015
roll W × L
300 mm × 4 m
pore size
0.2 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Western blotting
- Southern blotting
- Northern blotting
Features and Benefits
- High binding capacity.
- Versatile, robust and excellent signal-to-noise ratios.
- Optimal for applications when stripping and reprobing is required.
- Suitable for proteins of a wide range of molecular weights.
Storage and Stability
Legal Information
Storage Class Code
4.1B - Flammable solid hazardous materials
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
This page shows and discusses three protocols for stripping and reprobing a western blot membrane.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service