Skip to Content
Merck
All Photos(1)

Key Documents

GE10600005

Amersham Protran® Western blotting membranes, nitrocellulose

pore size 0.1 μm, roll W × L 200 mm × 4 m, pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22
Pricing and availability is not currently available.

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600005

roll W × L

200 mm × 4 m

pore size

0.1 μm pore size

Looking for similar products? Visit Product Comparison Guide

General description

A versatile nitrocellulose blotting membrane with 0.1 µm pore size. Amersham Protran offers excellent sensitivity, resolution and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western blotting systems.

Application

Amersham Protran® Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Excellent signal-to-noise ratios and low background
  • Compatible with various labeling and detection systems
  • Offers a high binding capacity of small proteins (< Mr 10 000)
  • Reduces "blow-through" risk
  • Bound proteins have an extended shelf life due to the proprietary nitrocellulose formula
  • Precut sizes are available for laboratory application

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service