Skip to Content
Merck
All Photos(1)

Key Documents

G9774

Sigma-Aldrich

Guinea pig serum

Synonym(s):

Serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

guinea pig

Quality Level

sterility

non-sterile

contains

preservative

shipped in

dry ice

storage temp.

−20°C

General description

Guinea pig serum is pooled from normal donor herd.

Application

Guinea pig serum has been used as a neural cell culture medium (N2B27) supplement during immunosurgery to isolate inner cell masses. It has also been used as a component of sterile phosphate-buffered saline (PBS) in antibody assays.
Guinea pig serum is suitable blocking agent and negative control in immunoassays.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced serum concentration is permissive for increased in vitro endocrine differentiation from murine embryonic stem cells
Vincent RK and Odorico JS
Differentiation, 78(1), 24-34 (2009)
Pneumonitis in Syrian golden hamsters (Mesocricetus auratus) infected with Rio Mamore virus (family Bunyaviridae, genus Hantavirus)
ML Milazzo, et al.
Virus Research, 191, 39-44 (2014)
Mary Louise Milazzo et al.
Virus research, 191, 39-44 (2014-07-30)
Rio Mamoré virus is an etiological agent of hantavirus pulmonary syndrome in South America. The purpose of this study was to determine whether Rio Mamoré virus strain HTN-007 in Syrian golden hamsters is pathogenic. None of 37 adult hamsters infected
Robert V House et al.
PloS one, 17(6), e0267796-e0267796 (2022-06-04)
The current global COVID-19 pandemic caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has resulted in a public health crisis with more than 168 million cases reported globally and more than 4.5 million deaths at the time of writing.
Andreas Postlmayr et al.
Development (Cambridge, England), 147(11) (2020-05-23)
We previously identified the cyclin dependent kinase Cdk8 as a putative silencing factor for Xist To investigate its role in X inactivation, we engineered a Cdk8 mutation in mouse embryonic stem cells (ESCs) carrying an inducible system for studying Xist

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service