Skip to Content
Merck
All Photos(1)

Key Documents

G9510

Sigma-Aldrich

Glutaryl coenzyme A lithium salt

≥90%

Synonym(s):

Glutaryl CoA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H42N7O19P3S
CAS Number:
Molecular Weight:
881.63
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Quality Level

Assay

≥90%

storage temp.

−20°C

SMILES string

[Li].CC(C)(COP(O)(=O)OP(O)(=O)OCC1OC(C(O)C1OP(O)(O)=O)n2cnc3c(N)ncnc23)C(O)C(=O)NCCC(=O)NCCSC(=O)CCCC(O)=O

InChI

1S/C26H42N7O19P3S/c1-26(2,21(39)24(40)29-7-6-15(34)28-8-9-56-17(37)5-3-4-16(35)36)11-49-55(46,47)52-54(44,45)48-10-14-20(51-53(41,42)43)19(38)25(50-14)33-13-32-18-22(27)30-12-31-23(18)33/h12-14,19-21,25,38-39H,3-11H2,1-2H3,(H,28,34)(H,29,40)(H,35,36)(H,44,45)(H,46,47)(H2,27,30,31)(H2,41,42,43)/t14-,19-,20-,21?,25-/m1/s1

InChI key

SYKWLIJQEHRDNH-KRPIADGTSA-N

General description

Glutaryl coenzyme A (Glutaryl CoA) is an intermediate in the mitochondrial oxidation of lysine, hydroxylysine and tryptophan.

Application

Glutaryl coenzyme A lithium salt has been used:
  • in comparative study of acylomes of β-Hydroxy β-methylglutaryl-CoA (HMG-CoA) and glutaryl-CoA by quantitative proteomics
  • as a component of the assay buffer for in vitro biosensor activity experiments for FapR-NLuc proteins
  • to test its effect on pyruvate kinase activity inhibition in a549 lysate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yipeng Du et al.
Bioconjugate chemistry, 30(3), 826-832 (2019-01-11)
Malonyl-CoA is one of the key metabolic intermediates in fatty acid metabolism as well as a key player in protein post-translational modifications. Detection of malonyl-CoA in live cells is challenging because of the lack of effective measuring tools. Here we
Kinetic mechanism of glutaryl-CoA dehydrogenase
Rao K S, et al.
Biochemistry, 45(51), 15853-15861 (2006)
Simon Wischgoll et al.
Biochemistry, 49(25), 5350-5357 (2010-05-22)
Glutaryl-coenzyme A dehydrogenases (GDHs) involved in amino acid degradation were thought to catalyze both the dehydrogenation and decarboxylation of glutaryl-coenzyme A to crotonyl-coenzyme A and CO(2). Recently, a structurally related but nondecarboxylating, glutaconyl-coenzyme A-forming GDH was characterized in the obligately
Rhushikesh A Kulkarni et al.
Cell chemical biology, 24(2), 231-242 (2017-02-07)
Non-enzymatic protein modification driven by thioester reactivity is thought to play a major role in the establishment of cellular lysine acylation. However, the specific protein targets of this process are largely unknown. Here we report an experimental strategy to investigate
Discovering targets of non-enzymatic acylation by thioester reactivity profiling
Kulkarni RA, et al.
Cell Chemical Biology, 24(2), 231-242 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service