Skip to Content
Merck
All Photos(1)

Key Documents

G8923

Sigma-Aldrich

GANT58

≥98% (HPLC)

Synonym(s):

2,3,4,5-Tetra(4-pyridyl)thiophene, 4,4′,4′′,4′′′-Thiene-2,3,4,5-tetrayltetrapyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16N4S
CAS Number:
Molecular Weight:
392.48
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

color

white to pale yellow

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

c1cc(ccn1)-c2sc(-c3ccncc3)c(-c4ccncc4)c2-c5ccncc5

InChI

1S/C24H16N4S/c1-9-25-10-2-17(1)21-22(18-3-11-26-12-4-18)24(20-7-15-28-16-8-20)29-23(21)19-5-13-27-14-6-19/h1-16H

InChI key

USWLOKMMUTWFMD-UHFFFAOYSA-N

Biochem/physiol Actions

GANT58 is a Hedgehog signaling pathway inhibitor. GANT58 inhibits the hedgehog signaling pathway downstream of Smo. GANT58 inhibits Gli transcriptional activity, and reduces cellular levels of the Hh pathway components Gli1, Shh, and patched.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Ciucci et al.
PloS one, 8(3), e60145-e60145 (2013-04-05)
Recent evidence links aberrant activation of Hedgehog (Hh) signaling with the pathogenesis of several cancers including medulloblastoma, glioblastoma, melanoma as well as pancreas, colorectal, and prostate carcinomas. Here we investigated the role of the transcription factor Gli1 in ovarian cancer.
Joseph Vanderburgh et al.
ACS nano, 14(1), 311-327 (2020-01-03)
Breast cancer patients are at high risk for bone metastasis. Metastatic bone disease is a major clinical problem that leads to a reduction in mobility, increased risk of pathologic fracture, severe bone pain, and other skeletal-related events. The transcription factor
Helen O Oladapo et al.
Cancer letters, 411, 136-149 (2017-10-03)
Activation of the Hedgehog (Hh) pathway effector GLI1 is linked to tumorigenesis and invasiveness in a number of cancers, with targeting of GLI1 by small molecule antagonists shown to be effective. We profiled a collection of GLI antagonists possessing distinct
Ferdinando Maria Milazzo et al.
Frontiers in oncology, 9, 1534-1534 (2020-02-11)
Targeted therapy using monoclonal antibodies conjugated to toxins is gaining space in the treatment of cancer. Here, we report the anti-tumor effect of a new antibody drug conjugate (ADC) delivering a HDAC inhibitor to ErbB2+ solid tumors. Trastuzumab was partially
Se-Young Cho et al.
Fish & shellfish immunology, 88, 244-253 (2019-02-26)
Seasonal temperature has a major influence on the infectivity of pathogens and the host immune system. Viral hemorrhagic septicemia virus (VHSV) is one such pathogen that only causes the mortality of fish at low temperatures. This study aims to discover

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service