Skip to Content
Merck
All Photos(1)

Key Documents

G8903

Sigma-Aldrich

Ghrelin rat

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

form

lyophilized powder

Quality Level

color

white

solubility

1% acetic acid: 1 mg/mL
water: 1 mg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Ghrl(59301)

Amino Acid Sequence

Gly-Ser-Ser(O-n-octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg

General description

Ghrelin is a gut-brain orexigenic peptide that functions in hunger signaling. This peptide is prominently synthesized by stomach and transported to brain. However, a specific subset of hypothalamic neurons also produces this peptide. It is expressed in gastrointestinal tract, hypothalamic arcuate nucleus, pituitary, kidney, liver, heart, lung and adipose tissue This peptide is composed of 28 amino acids, and its Ser3 residue is N-octanoylated.

Application

Ghrelin rat has been used for intraperitoneal injections, and microinjections in rats to study its effects on celiac disease, and passive avoidance learning (PAV) and spatial learning, respectively. It has also been used for intracerebroventricular injection in rats to study its influence on penicillin-induced epileptiform activity in rat.

Biochem/physiol Actions

Ghrelin acts as a ligand for G-protein coupled receptor, GHS-R1A (growth hormone secretagogue receptor), and controls energy utilization and feeding. This peptide is involved in enhancing the value of rewarding foods, drugs and alcohol. In rats with diabetes, the expression of ghrelin increased as a result of cachexia. In rats with ovariectomy, the expression of this peptide decreased in the endometrium. This peptide controls cognitive functions and has neuroprotective roles in neurodegenerative disorders. In cultured cortical neuronal networks, ghrelin increases the rate of synapse formation and activity development.
Endogenous ligand to the orphan growth-hormone secretagogue receptor that stimulates the release of growth hormone from the pituitary.

Physical form

Lyophilized from 0.1% aqueous TFA

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Funda Unsal et al.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University, 23(3), 363-370 (2014-07-01)
Ghrelin is a hormone which has effects on the secretion of growth hormone, the gastrointestinal system, the cardiovascular system, cell proliferation and the reproductive system. The aim of this study is to investigate structural changes in the uterine tissue and
Ali Aslan et al.
Neuropeptides, 43(4), 295-302 (2009-06-26)
Ghrelin, a gastric peptide with key action on food intake, has been recently recognized as a potential antiepileptic agent. In the present study, we investigated the involvement of nitric oxide in the effect of ghrelin on penicillin-induced epileptiform activity in
Irina I Stoyanova et al.
BMC neuroscience, 15, 49-49 (2014-04-20)
While ghrelin was initially related to appetite stimulation and growth hormone secretion, it also has a neuroprotective effect in neurodegenerative diseases and regulates cognitive function. The cellular basis of those processes is related to synaptic efficacy and plasticity. Previous studies
Mehmet F Sönmez et al.
Advances in clinical and experimental medicine : official organ Wroclaw Medical University, 24(3), 401-407 (2015-10-16)
Ghrelin is a hormone which has effects on the secretion of growth hormone, the gastrointestinal system, the cardiovascular system, cell proliferation and the reproductive system. This study focused on relative ghrelin and GHS-R1a gene expression in the stomach of long-term diabetic
Stewart Christie et al.
Peptides, 131, 170371-170371 (2020-07-14)
Ghrelin and anandamide (AEA) can regulate the sensitivity of gastric vagal afferents to stretch, an effect mediated via the transient receptor potential vanilloid 1 (TPRV1) channel. High fat diet (HFD)-induced obesity alters the modulatory effects of ghrelin and AEA on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service