Skip to Content
Merck
All Photos(1)

Key Documents

G6784

Sigma-Aldrich

ʟ-Glutamine–Penicillin–Streptomycin

solution, suitable for cell culture, BioReagent

Synonym(s):

Pen/Strep, Penicillin/Streptomycin 

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.76

Product Name

L-Glutamine–Penicillin–Streptomycin solution, L-glutamine 200 mM, streptomycin 10 mg/mL, penicillin 10,000 units, 0.1 μm filtered, BioReagent, suitable for cell culture

biological source

synthetic

Quality Level

description

stabilized

sterility

0.1 μm filtered

product line

BioReagent

form

liquid

composition

L-glutamine, 200 mM
penicillin, 10,000 units
streptomycin, 10 mg/mL

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes
protein synthesis | interferes

storage temp.

−20°C

Application

This antibiotic solution is used to reduce or eliminate bacterial contamination in primary cell cultures. It is recommended for us at 10 mL/L.

Biochem/physiol Actions

Mode of Action: The product acts in three ways: Glutamine is an essential energy source for most mammalian cells in culture due to its amino acid carbon skeleton. Penicillin inhibits bacterial cell wall synthesis. Streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome, causing miscoding.

Antimicrobial spectrum: This product acts against both gram-negative and gram-positive bacteria.

Components

This solution is formulated to contain 200 mM L-glutamine, 10,000 units of Penicillin and 10 mg/mL of streptomycin, solubilized in a proprietary buffer.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengliang L Wu et al.
Glycobiology, 30(7), 454-462 (2020-01-04)
Cells are covered with glycans. The expression and distribution of specific glycans on the surface of a cell are important for various cellular functions. Imaging these glycans is essential to aid elucidation of their biological roles. Here, utilizing methods of
Nikoletta Sztán et al.
Reproduction, fertility, and development, 29(11), 2206-2216 (2017-03-28)
The conservation of genetic resources of avian species has become increasingly important over the past decade. The aim of the present study was to develop a genome preservation technique for the Hungarian goose Anser anser domestica. To this end, we
Miriam Gonzalez Gonzalez et al.
Journal of immunological methods, 457, 73-77 (2018-02-25)
Standard cell culturing on plastic plates (two dimensional (2D) cultures) does not represent the actual microenvironment where cells reside in tissues. The three dimensional (3D) systems, composed of extracellular matrix and/or pure amino acids which form a scaffold for cells
Kate M Herum et al.
Journal of the American Heart Association, 9(3), e013518-e013518 (2020-02-01)
Background Pressure overload of the heart occurs in patients with hypertension or valvular stenosis and induces cardiac fibrosis because of excessive production of extracellular matrix by activated cardiac fibroblasts. This initially provides essential mechanical support to the heart, but eventually
A Y Hafez et al.
Oncogenesis, 6(6), e349-e349 (2017-06-13)
Activation of cellular oncogenes as well as infection with tumor viruses can promote aberrant proliferation and activation of the host DNA damage response. Epstein-Barr virus (EBV) infection of primary human B cells induces a transient period of hyper-proliferation, but many

Articles

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Importance and uses of glutamine in hybridoma and mammalian cell culture

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service