Skip to Content
Merck
All Photos(1)

Key Documents

G6503

Sigma-Aldrich

4-Guanidinobutyric acid

≥98%, for peptide synthesis

Synonym(s):

γ-Guanidinobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH=C(NH2)NHCH2CH2CH2COOH
CAS Number:
Molecular Weight:
145.16
Beilstein:
1766447
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

product name

4-Guanidinobutyric acid, ≥98%

Quality Level

Assay

≥98%

form

powder or chunks

technique(s)

drug transporter assay: suitable

color

white

mp

280-282 °C

storage temp.

2-8°C

SMILES string

NC(=N)NCCCC(O)=O

InChI

1S/C5H11N3O2/c6-5(7)8-3-1-2-4(9)10/h1-3H2,(H,9,10)(H4,6,7,8)

InChI key

TUHVEAJXIMEOSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gamma-Guanidinobutyric acid (4-GBA) is used to study intestinal transport by the human proton-coupled amino acid transporter(s) (hPAT1). 4-GBA is used with other model compounds such as 1,6-diaminohexane, 1,6-diguanidinohexane, and 1,6-di(cyanoguanidino)hexane to study the biodegradability of the biocide polyhexamethylene biguanide (PHMB).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhipan Zhang et al.
The journal of physical chemistry. B, 109(46), 21818-21824 (2006-07-21)
Dye-sensitized solar cells based on nanocrystalline TiO(2) have been fabricated with an amphiphilic ruthenium sensitizer [Ru (4,4'-dicarboxylic acid-2,2'-bipyridine) (4,4'-bis(p-hexyloxystyryl)-2,2'-bipyridine)(NCS)(2)], coded as K-19, and 4-guanidinobutyric acid (GBA) as coadsorbent. The cells showed a approximately 50 mV increase in open-circuit voltage and
Guanidino compounds in hyperargininemia.
B Marescau et al.
Advances in experimental medicine and biology, 153, 427-434 (1982-01-01)
M Al Banchaabouchi et al.
Metabolism: clinical and experimental, 47(3), 355-361 (1998-03-21)
Renal failure is characterized by the retention of nitrogenous metabolites such as urea, creatinine (CTN) and other guanidino compounds (GCs), uric acid, and hippuric acid, which could be related to the clinical syndrome associated with renal insufficiency. A model of
Disturbed metabolism of guanidino compounds characterized by elevated excretion of beta-guanidinopropionic acid and gamma-guanidinobutyric acid--an effect of vigabatrin treatment?
A Schulze et al.
Journal of inherited metabolic disease, 21(3), 268-271 (1998-08-01)
C Cabella et al.
European journal of biochemistry, 268(4), 940-947 (2001-02-17)
Rat hepatocytes in culture take up [14C]-agmatine by both a high-affinity transport system [KM = 0.03 mM; Vmax = 30 pmol x min x (mg protein)-1] and a low-affinity system. The high-affinity system also transports putrescine, but not cationic amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service