Skip to Content
Merck
All Photos(1)

Key Documents

G6501

Sigma-Aldrich

Glycerol phosphate disodium salt hydrate

isomeric mixture

Synonym(s):

Glycerophosphate disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7Na2O6P · xH2O
CAS Number:
Molecular Weight:
216.04 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

form

powder

SMILES string

O.[Na].OCC(CO)OP(O)(O)=O

InChI

1S/C3H9O6P.2Na.H2O/c4-1-3(2-5)9-10(6,7)8;;;/h3-5H,1-2H2,(H2,6,7,8);;;1H2/q;2*+1;/p-2

InChI key

ROPZSVKNEIIIDE-UHFFFAOYSA-L

Related Categories

Application

Evaluated as phosphorus source in the cultivation of Pichia pastoris. Used along with chitosan to prepare biodegradable hydrogels.
Glycerol phosphate disodium salt was used in a novel microplate-based chromatin immunoprecipitation method to immunoprecipitate DNA from rat mesangial cells in a 96-well format.

Physical form

Usually contains 50% β-isomer and 50% rac-α-isomer

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David P Paulson et al.
The Laryngoscope, 118(4), 706-711 (2008-01-10)
Our goal is to develop a novel drug delivery system that can potentially improve clinical outcomes compared to current methods of dosing drugs such as dexamethasone or gentamicin. This system focuses on a single local application to the inner ear
Wenhui Zhang et al.
Applied microbiology and biotechnology, 72(1), 139-144 (2006-01-31)
Pichia pastoris has emerged as a commercially important yeast for the production of a vast majority of recombinant therapeutic proteins and vaccines. The organism can be grown to very high cell densities using a defined basal salts media (BSM). However
Yangling Hu et al.
British journal of cancer, 123(11), 1644-1655 (2020-09-17)
Microcalcification is one of the most reliable clinical features of the malignancy risk of breast cancer, and it is associated with enhanced tumour aggressiveness and poor prognosis. However, its underlying molecular mechanism remains unclear. Clinical data were retrieved to analyse
Yannan Tang et al.
Assay and drug development technologies, 17(6), 267-274 (2019-08-14)
Phospholipid biosynthesis begins with the acylation of glycerol 3-phosphate (G3P). In most Gram-positive bacteria including many pathogens, a membrane protein called PlsY is the only acyltransferase that catalyzes this essential step, making it a potential target for the development of
Steve Flanagin et al.
Nucleic acids research, 36(3), e17-e17 (2008-01-22)
The chromatin immunoprecipitation (ChIP) assay is a major tool in the study of genomic processes in vivo. This and other methods are revealing that control of gene expression, cell division and DNA repair involves multiple proteins and great number of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service