Skip to Content
Merck
All Photos(1)

Key Documents

G6137

Sigma-Aldrich

Glutathione Peroxidase from bovine erythrocytes

lyophilized powder, ≥300 units/mg protein

Synonym(s):

GSH-Px, Glutathione:hydrogen-peroxide oxido-reductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bovine erythrocytes

Quality Level

form

lyophilized powder

specific activity

≥300 units/mg protein

mol wt

84.5 kDa

composition

Protein, 10-30% modified Warburg-Christian

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Glutathione peroxidase is an antioxidant enzyme that contains selenium. It is present in the glandular epithelium of human endometrium.

Application

Glutathione Peroxidase from bovine erythrocytes has been used as an oxygen radical scavenger to study its effect on cytotoxicity of 1,3-dilinoleoylglycerol (DLG) against E1A-3Y1 cells.
Glutathione peroxidase from bovine erythrocytes was used as a positive control in cloning and characterization of full-length cDNAs encoding two glutathione peroxidases (GpXs) from Globodera rostochiensis. It was used for the determination of glutathione peroxidase activity in human milk.

Biochem/physiol Actions

Glutathione peroxidase helps to reduce (peroxides) H2O2 to water and lipid peroxides to lipid alcohols.
Glutathione peroxidase is an enzyme which reduced lipid hydroperoxides into their corresponding alcohols. It also reduces free hydrogen peroxide in to water. In vivo it is responsible for protecting hemoglobin from oxidative breakdown.
Protects cells against oxidative damage by catalyzing the reduction of hydrogen peroxide in the presence of reducing agent glutathione. In cellular membranes it may induce lipid peroxidation through the reduction of hydrogen peroxide or polyunsaturated fatty acid hydroperoxides.

Unit Definition

One unit will catalyze the oxidation by H2O2 of 1.0 μmole of reduced glutathione to oxidized glutathione per min at pH 7.0 at 25 °C.

Physical form

Lyophilized powder containing 25% sucrose and 2.5% dithiothreitol with sodium phosphate buffer salts

Other Notes

Note: At the reported pH optimum of 8.8, we have found the activity to be approx. 10 times that at pH 7.0. However, to remain consistent with literature and avoid complications arising from non-enzymatic oxidation of GSH, our unit is defined at pH 7.0.

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactive oxygen species, oxidative stress, and vascular biology in hypertension
Comprehensive hypertension, 337-344 (2007)
Yue Wang et al.
Analytical chemistry, 92(2), 1997-2004 (2019-12-21)
Solid evidence confirms that glutathione peroxidase (GPx) is a kind of vital protease in the first-line antioxidant defense system and participates in regulation of redox homeostasis as well as the pentose phosphate pathway. However, the current methods cannot achieve real-time
J T Jones et al.
Gene, 324, 47-54 (2003-12-25)
We report the cloning and characterisation of full-length DNAs complementary to RNA (cDNAs) encoding two glutathione peroxidases (GpXs) from a plant parasitic nematode, the potato cyst nematode (PCN) Globodera rostochiensis. One protein has a functional signal peptide that targets the
Angeles Torres et al.
Die Nahrung, 47(6), 430-433 (2004-01-20)
An analytical method for determining glutathione peroxidase (GPx) (EC 1.11.1.9) activity in whole blood has been adapted to human milk samples. The values obtained for precision (relative standard deviation: 8.4%), linearity and accuracy (recovery: 90.4%) indicate the adequacy of the
The role of oxidative stress in endometriosis
Handbook of Fertility, 273-281 (2015)

Articles

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Cellular oxidative stress is countered by enzymatic scavengers and antioxidant modulators against reactive oxygen species damage.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service