Skip to Content
Merck
All Photos(1)

Key Documents

G5760

Sigma-Aldrich

Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides

Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret

Synonym(s):

G-6-P-DH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Leuconostoc mesenteroides)

Quality Level

type

Type XXIII

form

ammonium sulfate suspension

specific activity

550-1,100 units/mg protein (biuret)

mol wt

54 kDa

concentration

≥2.0 mg/mL Biuret

foreign activity

6-Phosphogluconic dehydrogenase, hexokinase, NADH oxidase and NADPH oxidase ≤0.005%
PGI ≤0.01%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Research area: Cell Signaling

Glucose-6-phosphate dehydrogenase (G6PD) is found in the cytoplasm of all cells. The G6PD enzyme is encoded by the Gd gene, located on the long arm of the X chromosome.Glucose-6-phosphate Dehydrogenase (G6PD) from Leuconostoc mesenteroides catalyzes the oxidation of glucose 6-phosphate in the presence of nicotinamide adenine dinucleotide (NAD+) or nicotinamide adenine dinucleotide phosphate (NADP+). It corresponds to a molecular weight of 54 kDa and exists as a homodimer.

Application

Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides has been used:

  • as a component of reaction mixture for assaying mannose- and glucose-6-phosphate activity
  • as a component of reaction mixture in the nevirapine inhibition studies in human hepatic microsomes
  • in the glucose-phosphorylating activity of chloroplast extracts
  • as a model to test the effect of seed protein fractions on enzyme protection during dehydration.
  • in assays for nicotinamide adenine dinucleotide and tissue pyridine nucleotides.

Biochem/physiol Actions

Glucose-6-phosphate dehydrogenase (G6PD) catalyzes the conversion of glucose-6-phosphate to 6-phosphogluconolacetone as the first step in the pentose phosphate pathway.
Glucose-6-phosphate dehydrogenase (G6PD) is a housekeeping enzyme that protects cells from damage caused by reactive oxygen species by supplying substrates that help prevent oxidative harm. G6PD from Leuconostoc mesenteroides catalyzes the oxidation of glucose 6-phosphate in the presence of nicotinamide adenine dinucleotide (NAD+) or nicotinamide adenine dinucleotide phosphate (NADP+) as the first step in the pentose phosphate pathway. Lysine 21 in the Glucose-6-phosphate Dehydrogenase (G6PD) is crucial for binding NAD+. G6PD uses NAD+ or NADP+ based on the catabolic or anabolic metabolic pathway.G6PD deficiency can lead to acute hemolysis, neonatal jaundice, or severe chronic non-spherocytic hemolytic anemia.

Unit Definition

One unit will oxidize 1.0 μmole of D-glucose 6-phosphate to 6-phospho-D-gluconate per min in the presence of NAD at pH 7.8 at 30 °C.

Physical form

Suspension in 2.7 M (NH4)2SO4 solution containing 42 mM Tris and 0.8 mM MgCl2

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucose-6-Phosphate Dehydrogenase Deficiency
Richardson SR and O'Malley GF
StatPearls [Internet] (2022)
In vitro studies: inhibition of nevirapine metabolism by nortriptyline in hepatic microsomes
Usach I and Peris JE
Bio-protocol, 5(19) (2014)
Oligomerization studies of Leuconostoc mesenteroides G6PD activity after SDS-PAGE and blotting
Ravera S, et al.
Molecular Biology, 44(3), 415-419 (2010)
A novel type of chloroplast stromal hexokinase is the major glucose-phosphorylating enzyme in the moss Physcomitrella patens
Olsson T, et al.
The Journal of Biological Chemistry, 278(45), 44439-44447 (2003)
A Modified Method for Detection of Post-Phoretic Activities in Mannose-(Mpi, EC 5.3. 1.8) and Glucose-(Gpi, EC 5.3. 1.9) 6-Phosphate Isomerases
Zasypkin MY, et al.
Russian Journal of Genetics, 37(5), 581-583 (2001)

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Protocols

To measure glucose-6-phosphate dehydrogenase activity, beta-nicotinamide adenine dinucleotide phosphate is used in a spectrophotometric rate determination assay at 340 nm.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service