Skip to Content
Merck
All Photos(1)

Key Documents

G3798

Sigma-Aldrich

10074-G5

≥98% (HPLC)

Synonym(s):

Biphenyl-2-yl-(7-nitrobenzo[1,2,5]oxadiazol-4-yl)amine, N-2-Biphenylyl-7-nitro-2,1,3-benzoxadiazol-4-amine, N-[1,1′-Biphenyl-2-yl]-7-nitro-2,1,3-Benzoxadiazol-4-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12N4O3
CAS Number:
Molecular Weight:
332.31
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

red

solubility

DMSO: >10 mg/mL

storage temp.

room temp

SMILES string

[O-][N+](=O)c1ccc(Nc2ccccc2-c3ccccc3)c4nonc14

InChI

1S/C18H12N4O3/c23-22(24)16-11-10-15(17-18(16)21-25-20-17)19-14-9-5-4-8-13(14)12-6-2-1-3-7-12/h1-11,19H

InChI key

KMJPYSQOCBYMCF-UHFFFAOYSA-N

Biochem/physiol Actions

10074-G5 is a c-Myc/Max interaction inhibitor. The c-Myc oncoprotein and its partner Max are intrinsically disordered (ID) monomers that undergo coupled folding and binding upon heterodimerization. 10074-G5, similarly to 10058-F4 (#F3680), specifically inhibits this interaction by binding to c-Myc, thus preventing C-Myc specific DNA binding and target genes regulation. 10074-G5 (2.8 microM) is slightly more potent that 10058-F4 (5.2 microM). It was discovered that 10074-G5 binds to a different specific binding site (region) of C-Myc than 10054-F4. Thus, the compound may become desirable for probing different interactions.

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alina Castell et al.
Scientific reports, 8(1), 10064-10064 (2018-07-04)
MYC is a key player in tumor development, but unfortunately no specific MYC-targeting drugs are clinically available. MYC is strictly dependent on heterodimerization with MAX for transcription activation. Aiming at targeting this interaction, we identified MYCMI-6 in a cell-based protein
Philipp Raffeiner et al.
Oncotarget, 5(19), 8869-8878 (2014-10-20)
The oncogenic bHLH-LZ transcription factor Myc forms binary complexes with its binding partner Max. These and other bHLH-LZ-based protein-protein interactions (PPI) in the Myc-Max network are essential for the physiological and oncogenic activities of Myc. We have generated a genetically
Quan Yang et al.
Frontiers in immunology, 12, 627072-627072 (2021-03-13)
The accumulation of myeloid-derived suppressor cells (MDSCs) is one of the major obstacles to achieve an appropriate anti-tumor immune response and successful tumor immunotherapy. MDSCs in tumor-bearing hosts are primarily polymorphonuclear (PMN-MDSCs). However, the mechanisms regulating the development of MDSCs
Udom Lao-On et al.
Biochimica et biophysica acta. Molecular basis of disease, 1866(3), 165656-165656 (2019-12-25)
Here we showed that the c-Myc oncogene is responsible for overexpression of pyruvate carboxylase (PC) in highly invasive MDA-MB-231 cells. Pharmacological inhibition of c-Myc activity with 10074-G5 compound, resulted in a marked reduction of PC mRNA and protein, concomitant with
Gabriella T Heller et al.
Science advances, 6(45) (2020-11-06)
Disordered proteins are challenging therapeutic targets, and no drug is currently in clinical use that modifies the properties of their monomeric states. Here, we identify a small molecule (10074-G5) capable of binding and sequestering the intrinsically disordered amyloid-β (Aβ) peptide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service