Skip to Content
Merck
All Photos(4)

Key Documents

G1277

Sigma-Aldrich

Glass beads, acid-washed

212-300 μm (50-70 U.S. sieve)

Synonym(s):

Glass beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102422
NACRES:
NB.22

particle size

212-300 μm (50-70 U.S. sieve)

Related Categories

Application

Acid-washed glass beads have been used:
  • for the extraction of nucleic acids from pathogens
  • for the preparation of Yarrowialipolytica cell extracts
  • in chromatin immunoprecipitation assay of rat aorta tissues

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method for correcting underestimation of enteric pathogen genome quantities in environmental samples
Pengbo L, et al.
Journal of Microbiology (2021)
Valeria Di Dato et al.
Scientific reports, 5, 12329-12329 (2015-07-21)
Diatoms are among the most diverse eukaryotic microorganisms on Earth, they are responsible for a large fraction of primary production in the oceans and can be found in different habitats. Pseudo-nitzschia are marine planktonic diatoms responsible for blooms in coastal
Nicholas M Briggs et al.
Nature communications, 9(1), 3827-3827 (2018-09-22)
Catalysts consisting of metal particles supported on reducible oxides exhibit promising activity and selectivity for a variety of current and emerging industrial processes. Enhanced catalytic activity can arise from direct contact between the support and the metal or from metal-induced
Valeria Di Dato et al.
Scientific reports, 10(1), 5374-5374 (2020-03-28)
Prostaglandins (PGs) are hormone-like mediators in many physiological and pathological processes that are present in all vertebrates, in some terrestrial and aquatic invertebrates, and have also been identified in some macroalgae. They have recently been reported also in marine microalgae
Thomas Dubois et al.
mBio, 11(4) (2020-08-21)
The crust is the outermost spore layer of most Bacillus strains devoid of an exosporium. This outermost layer, composed of both proteins and carbohydrates, plays a major role in the adhesion and spreading of spores into the environment. Recent studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service