Skip to Content
Merck
All Photos(1)

Key Documents

G1166

Sigma-Aldrich

Monoclonal Anti-Glutamic Acid Decarboxylase 65 antibody produced in mouse

clone GAD-6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-GAD 65

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GAD-6, monoclonal

form

buffered aqueous solution

species reactivity

mammals (and some lower vertebrates)

technique(s)

immunohistochemistry: 1:100-1:500
western blot: 1:1,000-1:5,000

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GAD2(2572)
mouse ... Gad2(14417)

Specificity

Recognizes the lower molecular weight isoform of the two GAD isoforms identified in brain, GAD 65. Can be used for immunochemical localization of GABA secreting neurons.

Immunogen

purified rat brain GAD 65.

Physical form

Solution in 0.01 M phosphate buffered saline, containing 0.04% sodium azide.

Preparation Note

Purified using Protein A chromatography.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stathis S Leondopulos et al.
Journal of neural engineering, 9(2), 026015-026015 (2012-03-01)
Slow wave oscillations in the brain are essential for coordinated network activity but have not been shown to self-organize in vitro. Here, the development of dissociated hippocampal neurons into an active network with oscillations on multi-electrode arrays was evaluated in
Han-Byeol Kim et al.
Cell death discovery, 6, 73-73 (2020-08-21)
Neonatal maternal separation (NMS), as an early-life stress (ELS), is a risk factor to develop emotional disorders. However, the exact mechanisms remain to be defined. In the present study, we investigated the mechanisms involved in developing emotional disorders caused by
Azadeh Yazdan-Shahmorad et al.
eLife, 7 (2018-05-29)
Brain stimulation modulates the excitability of neural circuits and drives neuroplasticity. While the local effects of stimulation have been an active area of investigation, the effects on large-scale networks remain largely unexplored. We studied stimulation-induced changes in network dynamics in
Rossella De Cegli et al.
Nucleic acids research, 41(2), 711-726 (2012-11-28)
Gene expression profiles can be used to infer previously unknown transcriptional regulatory interaction among thousands of genes, via systems biology 'reverse engineering' approaches. We 'reverse engineered' an embryonic stem (ES)-specific transcriptional network from 171 gene expression profiles, measured in ES
Susanna Molas et al.
Nature neuroscience, 20(9), 1260-1268 (2017-07-18)
Novelty preference (NP) is an evolutionarily conserved, essential survival mechanism often dysregulated in neuropsychiatric disorders. NP is mediated by a motivational dopamine signal that increases in response to novel stimuli, thereby driving exploration. However, the mechanism by which once-novel stimuli

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service