Skip to Content
Merck
All Photos(3)

Key Documents

G0549

Sigma-Aldrich

Corosolic acid from Lagerstroemia speciosa

≥98% (HPLC)

Synonym(s):

Corosolic acid, (2α,3β)-2,3-Dihydroxyurs-12-en-28-oic acid, 2α-Hydroxyursolic acid, Colosolic acid, Corsolic acid, Glucosol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

methanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@H]1[C@H](O)C(C)(C)[C@@](CC[C@]2(C)[C@]3([H])CC=C4[C@@]2(C)CC[C@]5(C(O)=O)[C@@]4([H])[C@@H](C)[C@H](C)CC5)([H])[C@]3(C)C1

InChI

1S/C30H48O4/c1-17-10-13-30(25(33)34)15-14-28(6)19(23(30)18(17)2)8-9-22-27(5)16-20(31)24(32)26(3,4)21(27)11-12-29(22,28)7/h8,17-18,20-24,31-32H,9-16H2,1-7H3,(H,33,34)/t17-,18+,20-,21+,22-,23+,24+,27+,28-,29-,30+/m1/s1

InChI key

HFGSQOYIOKBQOW-ZSDYHTTISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Corosolic acid from Lagerstroemia speciosa is a pentacyclic triterpenoid found in a variety of plants, such as apples, basil, bilberries, cranberries, and prunes. It is a natural derivative of ursolic acid.

Application

Corosolic acid from Lagerstroemia speciosa has been used to study the inhibiting activity of ursolic acid and its derivative against colon cancer cells by causing degradation of β-catenins.

Biochem/physiol Actions

Corosolic acid from Lagerstroemia speciose shows a variety of biological activities such as anti-proliferation, apoptosis, and anti-carcinogenic. It exhibits an inhibiting effect on the post-challenge plasma glucose levels in humans. Corosolic acid, due to its ability to improve glucose metabolism, has been touted to be used in the management of Type-2 Diabetes. It improves hypertension, oxidative stress, and inflammation, thereby being a potential candidate to be used against atherosclerosis-related diseases.
Triterpene phytochemical found in medicinal herbal extracts. Possesses antiatherosclerotic, antihyperlipidemic, antioxidant, antiinflammatory, antifungal, antiviral and antineoplastic activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Yamaguchi et al.
Life sciences, 79(26), 2474-2479 (2006-09-09)
Corosolic acid (CRA), a constituent of banaba leaves, has been reported to have anti-inflammatory and hypoglycemic activities. The aim of this study was to determine the effects of CRA on metabolic risk factors including obesity, hypertension, hyperinsulinemia, hyperglycemia, and hyperlipidemia
Yanfeng Xu et al.
Cancer letters, 284(2), 229-237 (2009-05-22)
We investigated the response of human cervix adenocarcinoma HeLa cells to Corosolic acid (CRA) treatment. Our results showed that CRA significantly inhibited cell viability in both a dose- and a time-dependent manner. CRA treatment induced S cell-cycle arrest and caused
M Fukushima et al.
Diabetes research and clinical practice, 73(2), 174-177 (2006-03-22)
Corosolic acid (CRA) is a substance extracted from Lagerstroemia speciosa L. and has been reported to have biological activities in in vitro and experimental animal studies. In this study, 31 subjects were orally administered 10mg CRA or a placebo, on
Wenli Hou et al.
Phytotherapy research : PTR, 23(5), 614-618 (2008-12-25)
The potential antidiabetic activity of ethyl acetate extract of the leaves of Lagerstroemia speciosa (LSL) was investigated by alpha-amylase and alpha-glucosidase inhibition assay. Six pentacyclic triterpenes (oleanolic acid, arjunolic acid, asiatic acid, maslinic acid, corosolic acid and 23-hydroxyursolic acid) were
Myung Sun Lee et al.
Phytotherapy research : PTR, 24(1), 49-53 (2009-06-24)
Four ursane-type triterpenoids, corosolic acid (1), ilekudinol B (2), ursolic acid (3) and pomolic acid (4), were isolated from an EtOAc-soluble extract of the leaves of Weigela subsessilis. These bioactive compounds were evaluated for their glucose uptake activity and produced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service