Skip to Content
Merck
All Photos(2)

Key Documents

G0259

Sigma-Aldrich

D-Glucosaminic acid

≥98% (TLC)

Synonym(s):

2-Amino-2-deoxy-D-gluconic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO6
CAS Number:
Molecular Weight:
195.17
Beilstein:
1726033
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (TLC)

form

powder

optical activity

[α]20/D -15.0 to -14.0 °, c = 2.5% (w/v) in hydrochloric acid

technique(s)

thin layer chromatography (TLC): suitable

color

white to off-white

mp

235 °C

solubility

0.5 M HCl: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

N[C@H]([C@@H](O)[C@H](O)[C@H](O)CO)C(O)=O

InChI

1S/C6H13NO6/c7-3(6(12)13)5(11)4(10)2(9)1-8/h2-5,8-11H,1,7H2,(H,12,13)/t2-,3-,4-,5-/m1/s1

InChI key

UFYKDFXCZBTLOO-TXICZTDVSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beata Liberek et al.
Carbohydrate research, 340(11), 1876-1884 (2005-06-28)
Mono- and di-N-alkylated derivatives of 1,3,4,6-tetra-O-acetyl-2-amino-2-deoxy-beta-D-glucose (alkyl=methyl, ethyl, propyl, butyl, pentyl, hexyl, benzyl) were synthesised by the reductive alkylation of per-O-acetyl-d-glucosamine. (N-ethyl, N-propyl, N-butyl, N-pentyl and N-hexyl)-1,3,4,6-tetra-O-acetyl-2-amino-2-deoxy-beta-D-glucoses were deacetylated in order to attempt an enzymatic phosphorylation. All products were characterised
D Horton et al.
Carbohydrate research, 193, 49-60 (1989-10-31)
Acetylation of 2-amino-2-deoxy-D-gluconic acid (1) with acetyl chloride-pyridine gave 2,3-unsaturated six- and five-membered lactones (2 and 3). Their benzoylated analogs (4 and 5) were obtained by benzoylation of 1 with benzoyl chloride-pyridine. Reaction of 1 with hot acetic anhydride-sodium acetate
N-methyl-1-glucosaminic acid.
M L WOLFROM et al.
Journal of the American Chemical Society, 68(11), 2343-2345 (1946-11-01)
E T Zavras et al.
The Journal of parasitology, 70(6), 937-944 (1984-12-01)
It was shown previously that worm-conditioned saline (WCS) prepared from crowded 10-day-old H. diminuta inhibited the incorporation of 3H-thymidine into DNA in the anterior regions of uncrowded worms and that the inhibition was partially accounted for by succinate and acetate
Kenji Sasaki et al.
Bioorganic & medicinal chemistry, 12(6), 1367-1375 (2004-03-17)
Biological activity of N-acetyl-6-sulfo-beta-d-glucosaminides (6-sulfo-GlcNAc 1) having a structural homology to N-acetylneuraminic acid (Neu5Ac 2) and 2-deoxy-2,3-dehydro-N-acetylneuraminic acid (Neu5Ac2en 3) was examined in terms of inhibitory activity against influenza virus sialidase (influenza, A/Memphis/1/71 H3N2). pNP 6-Sulfo-GlcNAc 1a was proved to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service