Skip to Content
Merck
All Photos(1)

Key Documents

FLSAR

Sigma-Aldrich

Somatic cell ATP releasing reagent

for ATP determination

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.51

Quality Level

usage

sufficient for ~1000 assays

packaging

vial of 10 mL concentrate

storage temp.

−20°C

Application

Somatic cell ATP releasing reagent has been used:
  • for the measurement of adenosine triphosphate (ATP) in tissues and cultured cells
  • to measure ATP content in oocytes
  • to lyse the cells to measure ATP and adenosine diphosphate (ADP) content

Somatic cell ATP releasing reagent is an ATP determination reagent used in application such as bioluminescent determination of ATP in somatic cells. Use with kits such as Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit (FLASC).

Reconstitution

Reconstitute with sterile water.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ugochukwu Nnodu et al.
Journal of applied toxicology : JAT, 28(8), 1016-1020 (2008-07-16)
Pentachlorophenol (PCP) is used as a wood preservative and is found in human blood and urine. PCP causes significant decreases in the tumor-killing (lytic) function of human natural killer (NK) cells, a critical immune defense. The current study examined the
Supplementation of maturation medium with L-carnitine improves cryo-tolerance of bovine in vitro matured oocytes
Chankitisakul, Vibuntita et al.
Theriogenology, 79(4), 590-598 (2013)
Prolonged exposure to insulin suppresses mitochondrial production in primary hepatocytes
Liu H Y, et al.
Test, 284(21), 14087-14095 (2009)
Hydrogen sulfide improves cardiomyocyte function in a cardiac arrest model
Garcia NA, et al.
Annals of Transplantation : Quarterly of the Polish Transplantation Society, 22(4), 285-285 (2017)
Glutamate dehydrogenase (GDH) regulates bioenergetics and redox homeostasis in human glioma
Zhang J, et al.
Testing, 5(4), 1-12 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service