Skip to Content
Merck
All Photos(1)

Key Documents

FLE250

Sigma-Aldrich

Firefly Lantern Extract

for ATP determination

Synonym(s):

FLE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.81

form

solution

packaging

pkg of 250 mg dried lanterns

technique(s)

toxicology assay: suitable

storage temp.

−20°C

General description

The bioluminescent organism, Photinus pyralis firefly is the source of firefly lantern extract. The luciferase enzyme is 60 kDa enzyme which lacks prosthetic group and bound metal in its structure. The N-terminal harbors residues crucial for the bioluminescent activity and the C-terminal possess peroxisome targeting motif.

Application

Fireflies have been used in several studies of bioluminescence. In particular they may be used in assays to determine toxicity pathways because bioluminescence can quantitatively measure a wide range of biological activities.
Firefly Lantern Extract has been used in the detection of ATP concentrations by bioluminescence flash analysis in seawater samples, bioluminescence assay in river sediments samples, marine sediments and in quantifying extracellular ATP in red blood cells.

Biochem/physiol Actions

Firefly Lantern Extract containing luciferase catalyzes oxidative decarboxylation of luciferin (LH2) in the presence of ATP, oxygen and Mg2+ to yield a light-emitter oxyluciferin.

Other Notes

Crude luciferin and luciferase

Reconstitution

After reconstitution with 25 mL of water, vial will contain 0.06 M potassium phosphate, pH 7.4, and 0.02 M MgSO4.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Involvement of acetylcholinesterase and protein kinase C in the protective effect of caffeine against beta-amyloid-induced alterations in red blood cells
Carelli-Alinovi C, et al.
Biochimie, 121, 52-59 (2016)
Distribution patterns of organic pollutants and microbial processes in marine sediments across a gradient of anthropogenic impact
Zoppini A, et al.
Environmental Pollution (Barking, Essex : 1987), 242, 1860-1870 (2018)
Concentrations and uptake of dissolved organic phosphorus compounds in the Baltic Sea
Nausch M, et al.
Frontiers in Marine Science, 5(386) (2018)
Firefly bioluminescence: a mechanistic approach of luciferase catalyzed reactions
Marques SM, et al.
IUBMB Life, 61(1), 6-17 (2009)
Microbial responses to polycyclic aromatic hydrocarbon contamination in temporary river sediments: Experimental insights
Zoppini A, et al.
The Science of the Total Environment, 541, 1364-1371 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service