Skip to Content
Merck
All Photos(1)

Key Documents

F8927

Sigma-Aldrich

Flupirtine maleate salt

≥98% (HPLC)

Synonym(s):

2-Amino-6-[[(4-fluorophenyl)methyl]amino]-3-pyridinyl]-carbamic acid ethyl ester maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17FN4O2 · C4H4O4
CAS Number:
Molecular Weight:
420.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble >20 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O.CCOC(=O)Nc1ccc(NCc2ccc(F)cc2)nc1N

InChI

1S/C15H17FN4O2.C4H4O4/c1-2-22-15(21)19-12-7-8-13(20-14(12)17)18-9-10-3-5-11(16)6-4-10;5-3(6)1-2-4(7)8/h3-8H,2,9H2,1H3,(H,19,21)(H3,17,18,20);1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

DPYIXBFZUMCMJM-BTJKTKAUSA-N

Biochem/physiol Actions

Flupirtine is commercially available as maleate salt. It exists in two polymorphs : flupirtine maleate A and B. Flupirtine is useful in treating muscular spasm, muscle tension and muscle stiffness. It is known to be effective in relieving back pain. Along with cytoprotection, flupirtine also offers protection against neurodegenerative disorders such as multiple sclerosis, amyotrophic lateral sclerosis, Alzheimer′s disease, Parkinson′s disease, Huntington′s chorea and AID (acquired immunodeficiency) associated encephalopathy. Flupirtine is found to be a potential drug for eye-related problems like maculopathy including diabetic retinopathy, retinitis pigmentosa and glaucoma. It is also proved to be helpful in preventing cardiac associated disorders such as myocardial ischemia and infarction, cerebral ischemia and infarction. Hepatitis is also prevented by the use of flupirtine.
Non-opioid analgesic; cytoprotective versus PrP fragment 106-126

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin C Michel et al.
British journal of clinical pharmacology, 73(5), 821-825 (2011-11-03)
To determine efficacy of the analgesic flupirtine in the treatment of overactive bladder syndrome in a proof-of-concept study. Double-blind, double-dummy, three-armed comparison of flupirtine extended release (400 mg/day, titrated to 600 mg/day), tolterodine extended release (4 mg/day) and placebo for
Fang Yu et al.
Epilepsy research, 97(1-2), 64-72 (2011-08-13)
Activation of KCNQ-channels has been shown to decrease or reduce the propagation of neuronal excitation in the immature central nervous system, and KCNQ activators represent a new class of anticonvulsant compounds. Their effectiveness has been demonstrated in many seizure models
Anton Kolosov et al.
Pain medicine (Malden, Mass.), 13(11), 1444-1456 (2012-10-20)
Current treatments for cancer pain are often inadequate, particularly when metastasis to bone is involved. The addition to the treatment regimen of another drug that has a complementary analgesic effect may increase the overall analgesia without the necessity to increase
Felicia Klinger et al.
British journal of pharmacology, 166(5), 1631-1642 (2011-12-23)
Flupirtine is a non-opioid analgesic that has been in clinical use for more than 20 years. It is characterized as a selective neuronal potassium channel opener (SNEPCO). Nevertheless, its mechanisms of action remain controversial and are the purpose of this
Syed Mohammed Naser et al.
Journal of the Indian Medical Association, 110(3), 158-160 (2012-10-04)
The study was conducted to evaluate the efficacy and safety of flupirtine maleate 100 mg thrice daily compared to tramadol hydrochloride 50 mg thrice daily as postoperative pain management for 5 days. A total of 113 postoperative patients were recruited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service