Skip to Content
Merck
All Photos(1)

Key Documents

F4165

Sigma-Aldrich

Ficin from fig tree latex

powder, ≥0.1 unit/mg solid

Synonym(s):

Debricin, Ficain, higueroxyl delabarre

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

fig tree (latex)

Quality Level

form

powder

specific activity

≥0.1 unit/mg solid

mol wt

23.8 kDa

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Extinction Coefficient: E1% = 21.0 (280 nm)
pI: 9.0

Biochem/physiol Actions

Ficin is classified as a thiol protease. It contains a single reactive cysteine at its active site. The amino acid homology of the active site is similar to that of papain. Ficin will cleave proteins at the carboxyl side of Gly, Ser, Thr, Met, Lys, Arg, Tyr, Ala, Asn, and Val. The reported Km for the chromogenic substrate pGlu-Phe-Leu-p-nitroanilide is 0.43 mM. Ficin is inhibited by iodoacetamide, iodoacetic acid, N-ethylmaleimide, mercuric chloride, DFP (diisopropyl fluorophosphate), TLCK (Na-p-Tosyl-lysine chloromethyl ketone), and TPCK (N-Tosyl-L-phenylalanine chloromethyl ketone). Ficin can be used to generate high yielding F (ab′)2 fragments from mouse IgG1.

Unit Definition

One unit will produce a ΔA280 of 1.0 per min at pH 7.0 at 37 °C when measuring TCA soluble products from casein in a final volume of 10 ml (1 cm light path).
The enzyme is soluble in 1 M potassium phosphate buffer, pH 7.0 (0.25 mg/ml), yielding a clear solution.

inhibitor

Product No.
Description
Pricing

substrate

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yufang Yang et al.
Scientific reports, 7, 43141-43141 (2017-02-23)
Ficin is classified as a sulfhydryl protease isolated from the latex of fig trees. In most cases, a particular enzyme fits a few types of substrate and catalyzes one type of reaction. In this investigation, we found sufficient proofs for
S Fadýloğlu
Die Nahrung, 45(2), 143-146 (2001-05-31)
Fig tree latex (ficin) was immobilized on Celite by adsorption. The free and immobilized ficin were utilized in the production of teleme (a Turkish milk product). After immobilization, the optimal temperature of ficin was shifted from 60 to 80 degrees
Daniel Janvier et al.
Transfusion, 53(9), 1948-1955 (2012-12-18)
Rh proteins and the Wr(b) antigen, which results from an interaction between Band 3 and glycophorin A, are the most common targets for warm immunoglobulin (Ig)G autoantibodies. Apart from autoanti-Di(b) , a scarce specificity, IgG warm autoantibodies specific for Band
Kamsagara Basavarajappa Devaraj et al.
Journal of agricultural and food chemistry, 56(23), 11417-11423 (2008-11-11)
Ficin (EC 3.4.22.3), a cysteine proteinase isolated from the latex of a Ficus tree, is known to occur in multiple forms. Although crude ficin is of considerable commercial importance, ficin as such has not been fully characterized. A major ficin
Robertson D Davenport et al.
Transfusion, 44(6), 849-852 (2004-05-26)
Cefotetan can cause severe immune hemolytic anemia that may persist long after the drug is discontinued. To study the binding of cefotetan to RBCs, patients who received cefotetan were followed and tested for the presence of antibody to cefotetan. Patients

Protocols

This procedure may be used for all Ficin products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service