F4042
Monoclonal ANTI-FLAG® M5 antibody produced in mouse
clone M5, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-ddddk, Anti-dykddddk
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
M5, monoclonal
form
buffered aqueous solution
purified by
using Protein A
species reactivity
all
concentration
2-5 mg/mL
technique(s)
western blot (chemiluminescent): 10 μg/mL
isotype
IgG1
immunogen sequence
DYKDDDDK
shipped in
dry ice
storage temp.
−20°C
Related Categories
General description
Monoclonal ANTI-FLAG® M5 is a mouse antibody that binds to N-terminal Met-FLAG fusion proteins. It is useful for detection of N-terminal Met-FLAG fusion proteins expressed in mammalian and Drosophilae cells.
Method of purification - Protein A
Method of purification - Protein A
Immunogen
FLAG; peptide sequence DYKDDDDK
Application
Binds the FLAG peptide only when it is located at the amino terminus preceded by a methionine. Binding is not Ca2+-dependent. Useful for detecting cytoplasmically expressed Met-FLAG fusion proteins in mammalian crude cell extracts, but not recommended for fusion proteins expressed in E. coli.
Monoclonal ANTI-FLAG® M5 antibody produced in mouse has been used in western blotting and immunofluorescence.
Physical form
Solution in 10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide (w/v)
Preparation Note
Dilute the antibody to 10 mg/mL in Tris buffered saline (TBS): 0.05 M Tris, 0.15 M NaCl, pH 7.4.
Other Notes
Antibody is not calcium dependent.
Legal Information
ANTI-FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chih-Chao Liang et al.
Nature communications, 7, 12124-12124 (2016-07-14)
The Fanconi anaemia (FA) pathway is important for the repair of DNA interstrand crosslinks (ICL). The FANCD2-FANCI complex is central to the pathway, and localizes to ICLs dependent on its monoubiquitination. It has remained elusive whether the complex is recruited
Franz Oswald et al.
Nucleic acids research, 44(10), 4703-4720 (2016-02-26)
The transcriptional shift from repression to activation of target genes is crucial for the fidelity of Notch responses through incompletely understood mechanisms that likely involve chromatin-based control. To activate silenced genes, repressive chromatin marks are removed and active marks must
CCAAT/enhancer-binding protein beta regulates the repression of type II collagen expression during the differentiation from proliferative to hypertrophic chondrocytes
Ushijima T, et al.
The Journal of Biological Chemistry, 289(5), 2852-2863 (2014)
Bader Almuzzaini et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(8), 2860-2873 (2016-04-30)
Actin and nuclear myosin 1 (NM1) are regulators of transcription and chromatin organization. Using a genome-wide approach, we report here that β-actin binds intergenic and genic regions across the mammalian genome, associated with both protein-coding and rRNA genes. Within the
DEF-1/ASAP1 is a GTPase-activating protein (GAP) for ARF1 that enhances cell motility through a GAP-dependent mechanism
Furman C, et al.
The Journal of Biological Chemistry, 277(10), 7962-7969 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service