Skip to Content
Merck
All Photos(1)

Key Documents

F3929

Sigma-Aldrich

Anti-Follicle-Stimulating Hormone Receptor antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-FSHR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 7-14 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FSHR(2492)
mouse ... Fshr(14309)
rat ... Fshr(25449)

General description

Follicle stimulating hormone receptor (FSHR) belongs to the G protein-coupled receptors family and is a glycoprotein. FSHR gene is mapped to human chromosome 2p16.3. FSHR comprises hydrophilic domain, hydrophobic membrane regions, C-terminal extracellular domain, and short intracellular domain.

Immunogen

synthetic peptide coresponding to the N-terminal extracellular domain of human follicle-stimulating hormone receptor, conjugated to KLH. The immunizing peptide has 24% identity and 29% similarity with the mouse gene.

Application

Anti-Follicle-Stimulating Hormone Receptor antibody produced in rabbit has also been used in immunohistochemistry in ovine and reproductive tissues. It has also been used in western blotting.

Biochem/physiol Actions

Follicle stimulating hormone receptor (FSHR) is located on Sertoli cells of the testes and the granulosa cells of ovary. The pituitary glycoprotein hormone, FSH, binds to FSHR and regulates the function of gonads and overall fertility. In Sertoli cells, FSHR-mediated cell proliferation determines the spermatogenic output. In ovarian granulosa cells, FSHR signaling mediates follicular growth and maturation of oocytes. Polymorphisms in the FSHR is implicated in premature ovarian insufficiency.

Physical form

Solution in phosphate buffered saline containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenling Huang et al.
Reproductive biology and endocrinology : RB&E, 17(1), 80-80 (2019-10-21)
Whether follicle-stimulating hormone receptor (FSHR) polymorphisms are implicated in premature ovarian insufficiency (POI) remains controversial. Thus, we performed this study to explore correlation between FSHR polymorphisms and POI in human beings. Literature retrieve was conducted in PubMed, Medline, Embase and
A T Grazul-Bilska et al.
Domestic animal endocrinology, 71, 106391-106391 (2019-11-16)
Corpus luteum (CL), a transient endocrine gland critical for reproductive cyclicity and pregnancy maintenance, is controlled by numerous regulatory factors. Although LH is widely recognized as the major regulator, other factors may also affect luteal functions. It has been demonstrated
Leslie L Heckert et al.
Recent progress in hormone research, 57, 129-148 (2002-05-23)
Results from experiments using mouse models suggest that the role of follicle-stimulating hormone (FSH) in spermatogenesis is the regulation of Sertoli cell proliferation and, ultimately, the size and spermatogenic capacity of the testis. The regulation of the expression of the
Lingyuan Fu et al.
Journal of assisted reproduction and genetics, 30(5), 717-721 (2013-03-29)
Polycystic ovary syndrome (PCOS) is a common complex genetic endocrinopathy. It has high heritability, and twin studies indicate that it is a complex polygenic disorder. Searching for major genes of PCOS is crucial to clarify its molecular pathogenesis. A previous
Jitu W George et al.
Biology of reproduction, 84(1), 7-17 (2010-08-27)
Follicle-stimulating hormone (FSH), a pituitary glycoprotein hormone, is an integral component of the endocrine axis that regulates gonadal function and fertility. To transmit its signal, FSH must bind to its receptor (FSHR) located on Sertoli cells of the testis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service