Skip to Content
Merck
All Photos(1)

Key Documents

F2293

Sigma-Aldrich

Follicle Stimulating Hormone from porcine pituitary

Synonym(s):

FSH, Follitropin, pFSH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
42020504
NACRES:
NA.32

form

powder

Quality Level

packaging

pkg of 50 unit per vial

technique(s)

cell culture | mammalian: suitable

solubility

H2O: soluble 50 mg/mL

UniProt accession no.

storage temp.

2-8°C

InChI

1S/C79H125N19O23S/c1-39(2)34-53(90-67(110)51(27-29-57(102)103)87-66(109)49(20-14-15-31-80)86-70(113)54(35-45-18-12-11-13-19-45)92-76(119)63(43(9)99)96-65(108)48(81)38-122)72(115)93-60(40(3)4)75(118)91-55(36-46-23-25-47(101)26-24-46)71(114)88-52(28-30-58(104)105)69(112)97-64(44(10)100)77(120)94-61(41(5)6)74(117)89-50(21-16-32-84-79(82)83)68(111)95-62(42(7)8)78(121)98-33-17-22-56(98)73(116)85-37-59(106)107/h11-13,18-19,23-26,39-44,48-56,60-64,99-101,122H,14-17,20-22,27-38,80-81H2,1-10H3,(H,85,116)(H,86,113)(H,87,109)(H,88,114)(H,89,117)(H,90,110)(H,91,118)(H,92,119)(H,93,115)(H,94,120)(H,95,111)(H,96,108)(H,97,112)(H,102,103)(H,104,105)(H,106,107)(H4,82,83,84)/t43-,44-,48+,49+,50+,51+,52+,53+,54+,55+,56+,60+,61+,62+,63+,64+/m1/s1

InChI key

RPJRYFRWJUYJGY-AYWRSWFDSA-N

Gene Information

human ... FSHR(2492)
pig ... FSHB(396895)

Looking for similar products? Visit Product Comparison Guide

General description

FSH (follicle stimulating hormone) protein is composed of two subunits- α and β, where α subunit has resemblance to LH, hCG and TSH, which are glycoprotein hormones. α subunit has one glycosylation site and β subunit has two.

Application

Follicle stimulating hormone from porcine pituitary has been used for the in vitro maturation, fertilization and co-culture of cumulus oocyte complex and the co-culture of primary germ and Sertoli cells obtained from adult rat testes.

Biochem/physiol Actions

FSH in involved in the growth of ovarian follicles and steroidogenesis of ovaries. FSHR (follicle stimulating hormone receptor) interacts with FSH in the ovaries, and determines the responsiveness to this hormone. Depressed levels of FSHR result in poor responsiveness and influence folliculogenesis. In Caucasian population, certain polymorphisms in this gene are more commonly found in patients with polycystic ovary syndrome (PCOS).
The α-chain is not active; biological specificity is attributed to the β-chain. Induces maturation of Graafian follicles of the ovary; promotes development of germinal cells in males; activates cytosolic tyrosine kinase.

Other Notes

Two-chain glycoprotein hormone

Unit Definition

One unit is the activity of 1 mg of Armour FSH Standard G-94 by the method of Steelman, S.L., and Pohley, F.M., Endocrinology, 53, 604 (1953).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Evecen et al.
Theriogenology, 75(9), 1647-1651 (2011-03-15)
There have not been successful and repeatable methods for in vitro embryo production in the dog. Up to date, only one blactocyst has been achieved on in vitro culture. Since reproductive physiology of the dog is different from that of
K W Park et al.
Biology of reproduction, 65(6), 1681-1685 (2001-11-22)
The in vitro developmental potential of porcine nuclear transfer (NT) embryos was evaluated. Oocytes were matured for 42-44 h, and metaphase II-oocytes were enucleated. Fetal fibroblasts infected with the enhanced green fluorescent protein (EGFP) gene were serum-starved for 3-5 days.
Wanakan Singhasena et al.
Reproductive biology and endocrinology : RB&E, 12, 86-86 (2014-09-03)
Polymorphisms at codons 307 and 680 are the most commonly encountered allelic variants of the follicle-stimulating hormone receptor (FSHR) gene. Studies in Caucasians suggest that certain FSHR variants are more common in women with polycystic ovary syndrome (PCOS) than normal
Michele Morelli et al.
Gynecologic oncology, 129(3), 448-451 (2013-04-06)
The objective of this study is to compare ovarian function and surgical outcomes between patients affected by benign uterine pathologies submitted to total laparoscopic hysterectomy (TLH) plus salpingectomy and women in which standard TLH with adnexal preservation was performed. We
Kathryn S Timberlake et al.
Fertility and sterility, 100(6), 1735-1739 (2013-09-24)
To assess whether blood type was associated with diminished ovarian reserve (DOR) (day-3 follicle-stimulating hormone level >10 IU/L), controlling for history of tobacco smoking, body mass index (BMI), history of endometriosis, ovarian surgery, previous pregnancy, and maternal age. Cross-sectional study.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service