Skip to Content
Merck
All Photos(1)

Key Documents

F0681

Sigma-Aldrich

Factor V human

aqueous glycerol solution, ≥25 units/mg protein

Synonym(s):

Labile factor, Proaccelerin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61
Pricing and availability is not currently available.

biological source

human

Quality Level

form

aqueous glycerol solution

specific activity

≥25 units/mg protein

technique(s)

ligand binding assay: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... F5(2153)

General description

Factor V is a single chain glycoprotein involved in the blood coagulation cascade. It is the procofactor of Va which accelerates the conversion of prothrombin to thrombin.

Application

Factor V is a key component in blood coagulation systems, where deficiencies can inhibit thrombin generation and affect hemostasis. It has been used in studies of Coagulopathic bleeding which occurs following injury and is a leading cause of in hospital deaths.

Linkage

View this factors role in the Coagulation Cascade.

Unit Definition

One unit is equivalent to the Factor V activity in 1.0 mL of normal human plasma at pH 7.4 at 37 °C.

Physical form

Aqueous solution containing 50% (v/v) glycerol

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Early coagulopathy in trauma patients: An on-scene and hospital admission study.
Floccard B., et al.
Injury (2010)
Eliza A Ruben et al.
Blood, 137(22), 3137-3144 (2021-03-09)
Coagulation factor V (fV) is the precursor of fVa, which, together with fXa, Ca2+, and phospholipids, defines the prothrombinase complex and activates prothrombin in the penultimate step of the coagulation cascade. We solved the cryogenic electron microscopy (cryo-EM) structures of
Sandro B Rizoli et al.
The Journal of trauma, 71(5 Suppl 1), S427-S434 (2011-12-17)
Coagulopathic bleeding is a leading cause of in-hospital death after injury. A recently proposed transfusion strategy calls for early and aggressive frozen plasma transfusion to bleeding trauma patients, thus addressing trauma-associated coagulopathy (TAC) by transfusing clotting factors (CFs). This strategy

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service