Skip to Content
Merck
All Photos(1)

Key Documents

F0679

SAFC

Fetal Bovine Serum

non-USA origin, γ-irradiated, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Synonym(s):

FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

biological source

bovine fetus

Quality Level

sterility

γ-irradiated
sterile-filtered

product line

FBS Classic

composition

hemoglobin, ≤25 mg/dL

origin

non-USA origin

availability

available only in EU
not available in Canada

technique(s)

cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

FBS remains a popular media supplement because it provides a wide array of functions in cell culture. FBS delivers nutrients, growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SFM) systems.

Application

FBS derived from clotted blood is the most widely used undefined supplement in eucaryotic, especially mammalian, cell culture. It is typically used with the classical media that were developed in the 1950s and 1960s. These include many popular media such as Minimum Essential Medium (MEM); Dulbecco′s Modified Eagles Media (DME, DMEM); Iscove′s Modified Eagles Medium (IMDM); M199 and Roswell Park Memorial Institute medium, RPMI-1640.
FBS is used in a broad range of cell culture applications. FBS provides many non-defined growth promoting and survival enhancing factor to cells in culture.

Analysis Note

  • Endotoxin and hemoglobin tested

Legal Information

Not available in USA or Canada. Please check with your local supplier regarding availability and import restrictions in your country.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Beatriz Vera-Pérez et al.
Journal of histotechnology, 42(4), 183-192 (2019-09-04)
Human adipose stem cells can differentiate into various mesodermic lineages, including adipogenic, osteogenic, chondrogenic, myogenic and endothelial pathways. In addition, these cells types possess immunomodulatory properties, potentially useful for autoimmune and autoinflammatory diseases. However, single-cell expanded clones have shown that

Related Content

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Explore our portfolio of fetal bovine serum (FBS) and determine the right FBS for your lab needs and budget. Request a serum sample.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service