EP5804719000
Eppendorf® Swing bucket rotor A-4-44
incl. conical form inserts
All Photos(1)
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Recommended Products
feature
to hold 4 x buckets for 2 x 50 mL conical tubes
incl. conical form inserts
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5804719000
parameter
5,000 max. RCF (4,500 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG