EP0030132882
Eppendorf® real-time PCR tube strips without caps
pack of 120 ea
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
Recommended Products
material
conical bottom
white tube
feature
graduations: no
packaging
pack of 120 ea
manufacturer/tradename
Eppendorf® 0030132882
technique(s)
PCR: suitable
capacity
0.1 mL
General description
- White wells for better reflection.
- High mechanical stability.
- Extremely thin walls for optimal heat transfer.
- Certified free from human DNA, DNase, RNase and PCR inhibitors.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG