EP0030122330
Eppendorf Tube®
amber tube, tube volume 5 mL, PCR clean, light protection, case of 200 ea (2 bags of 100 Tubes each)
About This Item
Recommended Products
material
conical bottom
polypropylene tube
transparent polypropylene
white polypropylene cap
sterility
sterile
feature
PCR clean light protection
conical shaped bottom with hinged lid
packaging
case of 200 ea (2 bags of 100 Tubes each)
manufacturer/tradename
Eppendorf® 0030122330
parameter
25,000 × g
tube volume
5 mL
color
amber tube
storage temp.
room temp
Looking for similar products? Visit Product Comparison Guide
General description
- Maximum safety and stability for centrifugation up to 25,000 × g
- Conical bottoms
- Easier processing of samples up to 5.0 mL
- Higher yields in DNA isolation, prep of mastermixes and buffers, and safe cell and tissue lysis
- Transparent polypropylene, free of plasticizers, biocides or mold release agents
- Hinged lid for minimized sample evaporation during storage and incubation in a wide range of temperatures from -86°C to 80°C
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service