Skip to Content

Fechas de fin de año: Facturación de pedidos hasta 13/12. Receso de Merck Argentina del 23/12 al 02/01.

Merck
All Photos(1)

Key Documents

EP0030073797

Eppendorf® epT.I.P.S.® G Reloads, PCR clean

volume (0.5-20 μL), L (46 mm), light gray, pack of 960 ea (10 trays × 96 tips)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
NACRES:
NB.02

Pricing and availability is not currently available. Contact Technical Service

material

gray (Tray)

sterility

non-sterile

packaging

pack of 960 ea (10 trays × 96 tips)

manufacturer/tradename

Eppendorf® 0030073797

technique(s)

PCR: suitable

L

(46 mm)

maximum volume

20 μL

volume

(0.5-20 μL)

color

light gray

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service