Skip to Content
Merck
All Photos(1)

Key Documents

EMU047641

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Brca1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGACTCCTTCCCAGGACAACTAGGTAGAAACAGAGGGCCTAAGGTGAACACTGTGCCTCCATTAGATAGTATGCAGCCTGGTGTCTGTCAGCAAAGTGTTCCTGTAAGTGATAAGTATCTTGAAATAAAAAAGCAGGAGGGTGAGGCTGTCTGTGCAGACTTCTCTCCATGTCTATTCTCAGACCATCTTGAGCAATCTATGAGTGGTAAGGTTTTTCAGGTTTGCTCTGAGACACCTGATGACCTGCTGGATGATGTTGAAATACAGGGACATACTAGCTTTGGTGAAGGTGACATAATGGAGAGATCTGCTGTCTTTAACGGAAGCATCCTGAGAAGGGAGTCCAGTAGGAGCCCTAGTCCTGTAACCCATGCATCGAAGTCTCAGAGTCTCCACAGAGCGTCTAGGAA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oreekha Amin et al.
BMC cancer, 15, 817-817 (2015-10-30)
Impairment of homologous recombination (HR) is found in close to 50 % of ovarian and breast cancer. Tumors with BRCA1 mutations show increased expression of the Insulin-like growth factor type 1 receptor (IGF-1R). We previously have shown that inhibition of
Nadire Duru et al.
Cancer letters, 369(1), 184-191 (2015-08-25)
Breast and lung cancer patients who are treated with radiotherapy often have severe side effects, including radiation-induced lung damage and secondary cancers. Activation of the NRF2 pathway is a well-known mechanism that protects cells against radiation induced oxidative stress, but
Samir Acharya et al.
PloS one, 9(8), e103819-e103819 (2014-08-02)
Fifteen percent of tumors utilize recombination-based alternative lengthening of telomeres (ALT) to maintain telomeres. The mechanisms underlying ALT are unclear but involve several proteins involved in homologous recombination including the BLM helicase, mutated in Bloom's syndrome, and the BRCA1 tumor
Min Peng et al.
The EMBO journal, 33(15), 1698-1712 (2014-06-27)
Several proteins in the BRCA-Fanconi anemia (FA) pathway, such as FANCJ, BRCA1, and FANCD2, interact with mismatch repair (MMR) pathway factors, but the significance of this link remains unknown. Unlike the BRCA-FA pathway, the MMR pathway is not essential for
Monique G P van der Wijst et al.
Molecular oncology, 9(7), 1259-1273 (2015-04-07)
Risk factors indicate the importance of oxidative stress during ovarian carcinogenesis. To tolerate oxidative stress, cells activate the transcription factor Nrf2 (Nfe2l2), the master regulator of antioxidant and cytoprotective genes. Indeed, for most cancers, hyperactivity of Nrf2 is observed, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service