Skip to Content
Merck
All Photos(1)

Key Documents

EMS0006

Sigma-Aldrich

Recombinant Trypsin

Proteomics Grade, lyophilized powder, recombinant, expressed in Pichia pastoris

Synonym(s):

Mass Spectrometry Trypsin, Proteomics grade Trypsin, rTrypsin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.26

recombinant

expressed in Pichia pastoris

Quality Level

grade

Proteomics Grade

form

lyophilized powder

specific activity

≥10,000 units/mg protein

suitability

suitable for
suitable for mass spectrometry

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

Trypsin is a major proteolytic enzyme, synthesized as a preproenzyme by pancreas and is stored as proenzyme trypsinogen in secretory granules. Trypsin belongs to the family of serine proteases that are characterized by the catalytic triad His57, Asp102 and Ser195. Trypsin is routinely used in proteomics research for peptide mapping and protein sequence work due to its highly specific cleavage resulting in a limited number of tryptic peptides. Trypsin is a pancreatic serine endoprotease which hydrolyzes peptide bonds specifically at the carboxyl side of arginine and lysine residues. The rate of hydrolysis is slower if an acidic residue is on either side of the cleavage site and cleavage may not occur if a proline residue is on the carboxyl side. The enzyme also exhibits esterase and amidase activities. Trypsin has an average molecular mass of 23.29 kDa and a pH optimum near 8.0. This product is prepared from recombinant trypsin, porcine sequence. It is naturally devoid of chymotryptic activity. This high-quality trypsin is suitable for proteomics use.

Biochem/physiol Actions

Trypsin plays an important role in the digestion of consumed protein and contributes to the activation of other proteolytic enzymes like chemotrypsin and elastase.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular characterization and gene expression of six trypsinogens in the flatfish Senegalese sole (Solea senegalensis Kaup) during larval development and in tissues
Manchado M, et al.
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology, 149(2), 334-344 (2008)
Structural properties of trypsin from cold-adapted fish, arabesque greenling (Pleurogrammus azonus)
Kanno G, et al.
European Food Research and Technology, 232(3), 381-388 (2011)
Autophagy receptor NDP52 alters DNA conformation to modulate RNA polymerase II transcription.
Dos Santos, et al.
Nature Communications, 14, 2855-2855 (2023)
Handbook of Proteolytic Enzymes (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service