Skip to Content
Merck
All Photos(1)

Key Documents

EHU152091

Sigma-Aldrich

MISSION® esiRNA

targeting human CASP8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCAAATGCAAACTGGATGATGACATGAACCTGCTGGATATTTTCATAGAGATGGAGAAGAGGGTCATCCTGGGAGAAGGAAAGTTGGACATCCTGAAAAGAGTCTGTGCCCAAATCAACAAGAGCCTGCTGAAGATAATCAACGACTATGAAGAATTCAGCAAAGAGAGAAGCAGCAGCCTTGAAGGAAGTCCTGATGAATTTTCAAATGACTTTGGACAAAGTTTACCAAATGAAAAGCAAACCTCGGGGATACTGTCTGATCATCAACAATCACAATTTTGCAAAAGCACGGGAGAAAGTGCCCAAACTTCACAGCATTAGGGACAGGAATGGAACACACTTGGATGCAGGGGCTTTGACCACGACCTTTGAAGAGCTTCATTTTGAGATCAAGCCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bang-Chuan Hu et al.
Theranostics, 10(25), 11479-11496 (2020-10-15)
Tubular damage initiated by inflammatory response and ischemic/hypoxic stress is a hallmark of septic acute kidney injury (AKI), albeit the molecular mechanism coupling the two events remains unclear. We investigated the intrinsic nature of tubular damage with respect to inflammatory/hypoxic
Bin Xu et al.
Oncology research, 25(7), 1161-1168 (2017-01-22)
Currently, multiple microRNAs (miRNAs) have been found to play vital roles in the pathogenesis of osteosarcoma. This study aimed to investigate the role of miR-21 in osteosarcoma. The level of miR-21 in 20 pairs of osteosarcoma and corresponding adjacent tissues
Hai-Yan Jia et al.
BMC molecular and cell biology, 20(1), 46-46 (2019-10-30)
It was reported that microRNA-21(miR-21) was differentially expressed in the keratinocytes of psoriasis patients, and it may influence the apoptosis and proliferation of cells. The role of lncRNA maternally expressed gene3 (MEG3), a competing endogenous RNAs of miR-21, in the
Jong-Heon Won et al.
Molecules (Basel, Switzerland), 23(12) (2018-12-16)
The natural product 23-hydroxyursolic acid (23-HUA) is a derivative of ursolic acid, which is known to induce cancer cell apoptosis. However, apoptotic effects and mechanisms of 23-HUA have not been well characterized yet. Herein, we investigated the molecular mechanisms of
Hui Chen et al.
The ocular surface, 18(4), 783-794 (2020-08-01)
Dry eye disease (DED) is a common and multifactor-induced autoimmune ocular surface disease. Environmental factors, such as desiccating stress (DS) and hyperosmolarity, affect the corneal epithelium to induce ocular surface inflammation in DED. We aimed to explore the potential mechanisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service