Skip to Content
Merck
All Photos(1)

Key Documents

EHU151981

Sigma-Aldrich

MISSION® esiRNA

targeting human HIF1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCAACATGGAAGGTATTGCACTGCACAGGCCACATTCACGTATATGATACCAACAGTAACCAACCTCAGTGTGGGTATAAGAAACCACCTATGACCTGCTTGGTGCTGATTTGTGAACCCATTCCTCACCCATCAAATATTGAAATTCCTTTAGATAGCAAGACTTTCCTCAGTCGACACAGCCTGGATATGAAATTTTCTTATTGTGATGAAAGAATTACCGAATTGATGGGATATGAGCCAGAAGAACTTTTAGGCCGCTCAATTTATGAATATTATCATGCTTTGGACTCTGATCATCTGACCAAAACTCATCATGATATGTTTACTAAAGGACAAGTCACCACAGGACAGTACAGGATGCTTGCCAAAAGAGGTGGATATGTCTGGGTTGAAACTCAAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Klaartje Somers et al.
Oncogene, 38(20), 3824-3842 (2019-01-24)
Survival rates for pediatric patients suffering from mixed lineage leukemia (MLL)-rearranged leukemia remain below 50% and more targeted, less toxic therapies are urgently needed. A screening method optimized to discover cytotoxic compounds selective for MLL-rearranged leukemia identified CCI-006 as a
Shu Lou et al.
Frontiers in cell and developmental biology, 8, 576-576 (2020-08-09)
Although genetic variants in autophagy pathway genes were associated with the risk of oral cancers and early development in embryos, their associations with non-syndromic cleft lip with or without cleft palate (NSCL/P) risk remained unclear. A two-stage case-control study (2,027
Qianqian Gao et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 74, 57-62 (2015-09-10)
A major cause of morbidity and mortality in cardiovascular disease is pathological cardiac hypertrophy. With an increase in the cellular surface area and upregulation of the atrial natriuretic peptide (ANP) gene, cardiac hypertrophy is a prominent feature of diabetic cardiomyopathy.
Yong Zhang et al.
Science translational medicine, 7(290), 290ra92-290ra92 (2015-06-05)
Whereas amphibians regenerate lost appendages spontaneously, mammals generally form scars over the injury site through the process of wound repair. The MRL mouse strain is an exception among mammals because it shows a spontaneous regenerative healing trait and so can
Olga Roche et al.
Nucleic acids research, 44(19), 9315-9330 (2016-11-02)
A wide range of diseases course with an unbalance between the consumption of oxygen by tissues and its supply. This situation triggers a transcriptional response, mediated by the hypoxia inducible factors (HIFs), that aims to restore oxygen homeostasis. Little is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service