Skip to Content
Merck
All Photos(1)

Key Documents

EHU140621

Sigma-Aldrich

MISSION® esiRNA

targeting human IRF5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGTCAGTTGGGCCTTCATAAACACTCACCTGGCTGGCTTTGCCTTCCAGGAGGAAGCTGGCTGAAGCAAGGGTGTGGAATTTTAAATGTGTGCACAGTCTGGAAAACTGTCAGAATCAGTTTTCCCATAAAAGGGTGGGCTAGCATTGCAGCTGCATTTGGGACCATTCAAATCTGTCACTCTCTTGTGTATATTCCTGTGCTATTAAATATATCAGGGCAGTGCATGTAAATCATCCTGATATATTTAATATATTTATTATATTGTCCCCCGAGGTGGGGACAGTGAGTGAGTTCTCTTAGTCCCCCCAGAGCTGGTTGTTAAAGAGCCTGGCACCTACCCGCTCTCACTTCATCTGTGTCATCTCTGCACACTCCAGCCCACTTTCTGCCTTCAGCCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matija Hedl et al.
Journal of immunology (Baltimore, Md. : 1950), 202(3), 920-930 (2018-12-30)
Common IFN regulatory factor 5 (IRF5) variants associated with multiple immune-mediated diseases are a major determinant of interindividual variability in pattern recognition receptor (PRR)-induced cytokines in macrophages. PRR-initiated pathways also contribute to bacterial clearance, and dysregulation of bacterial clearance can
Hongbin Cai et al.
Biochemical and biophysical research communications, 492(2), 192-198 (2017-08-19)
Ischemia-reperfusion injury (IRI) has been implicated in many pathological conditions, including cardiovascular diseases. Adhesion of leukocytes to the surface of endothelial cells has been considered as one of the principle steps in the pathological cascade of inflammatory tissue damage during
Jihyun Yang et al.
Journal of cellular physiology, 234(12), 23033-23042 (2019-05-28)
Bone-resorbing osteoclasts are differentiated from macrophages (MΦ) by M-CSF and RANKL. MΦ can be mainly classified into M1 and M2 MΦ, which are proinflammatory and anti-inflammatory, respectively, but little is known about their osteoclastogenic potential. Here, we investigated the osteoclastogenic
Kang Sun et al.
World journal of gastroenterology, 22(42), 9368-9377 (2016-11-30)
To investigate the role of interferon regulatory factor 5 (IRF5) in reversing polarization of lung macrophages during severe acute pancreatitis (SAP) A mouse SAP model was established by intraperitoneal (ip) injections of 20 μg/kg body weight caerulein. Pathological changes in
Jie Yan et al.
Journal of immunology (Baltimore, Md. : 1950), 205(4), 1024-1038 (2020-07-22)
Common IRF5 genetic risk variants associated with multiple immune-mediated diseases are a major determinant of interindividual variability in pattern-recognition receptor (PRR)-induced cytokines in myeloid cells. However, how myeloid cell-intrinsic IRF5 regulates the multiple distinct checkpoints mediating T cell outcomes in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service