Skip to Content
Merck
All Photos(1)

Key Documents

EHU138421

Sigma-Aldrich

MISSION® esiRNA

targeting human DHODH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCTGAAAGAGCAGGGCTTTGGCGGAGTCACAGATGCCATTGGAGCAGATCATCGGAGGTGAGGACAGCGTCTGACGGGAAGCCTGATCTGGAACCTTCCCAAGGACTCAGGCAAGCCTTTGTGGCTGGATCATGAGAGGAGGGACTCCATCTTGAGCCATGTCCCCCAGCCATGGCATGGCTGCACTGTAAACGCCAATCGGGGGGTCACCAGGATCAACCGCAGGCTTTCTTCAGTCCCTTGGTCAGACCATAAACTGCATTTTTGATTCTTTGTGGATTCAAACCCTAGGATCCATCAGTCTTGCAAGGACATTGAATATTAGGAGGAAAAAGTCATGGAAAAAATAAAGCCATTTAGAACCTGGGTTTCAACGCTAGCCCTTTCTGGTTTGCCATAGGCCCTGCCAAGATACTGCAGGTCCATCCAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xuemei Qiu et al.
International journal of oral science, 13(1), 3-3 (2021-01-30)
Oral squamous cell carcinoma (OSCC) become a heavy burden of public health, with approximately 300 000 newly diagnosed cases and 145 000 deaths worldwide per year. Nucleotide metabolism fuel DNA replication and RNA synthesis, which is indispensable for cell proliferation.
Mathura Subangari Dorasamy et al.
Journal of Cancer, 8(15), 3086-3098 (2017-09-21)
Dihydroorotate dehydrogenase (DHODH) is a rate-limiting enzyme in the
Deepti Mathur et al.
Cancer discovery, 7(4), 380-390 (2017-03-04)
Metabolic changes induced by oncogenic drivers of cancer contribute to tumor growth and are attractive targets for cancer treatment. Here, we found that increased growth of PTEN-mutant cells was dependent on glutamine flux through the de novo pyrimidine synthesis pathway
T He et al.
Oncogene, 33(27), 3538-3549 (2013-09-10)
Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) is a promising agent in selectively killing tumor cells. However, TRAIL monotherapy has not been successful as many cancer cells are resistant to TRAIL. Chemotherapeutic agents, such as doxorubicin have been shown to act

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service