Skip to Content
Merck
All Photos(1)

Key Documents

EHU133161

Sigma-Aldrich

MISSION® esiRNA

targeting human NR1H4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTGGACCATGAAGACCAGATTGCTTTGCTGAAAGGGTCTGCGGTTGAAGCTATGTTCCTTCGTTCAGCTGAGATTTTCAATAAGAAACTTCCGTCTGGGCATTCTGACCTATTGGAAGAAAGAATTCGAAATAGTGGTATCTCTGATGAATATATAACACCTATGTTTAGTTTTTATAAAAGTATTGGGGAACTGAAAATGACTCAAGAGGAGTATGCTCTGCTTACAGCAATTGTTATCCTGTCTCCAGATAGACAATACATAAAGGATAGAGAGGCAGTAGAGAAGCTTCAGGAGCCACTTCTTGATGTGCTACAAAAGTTGTGTAAGATTCACCAGCCTGAAAATCCTCAACACTTTGCCTGTCTCCTGGGTCGCCTGACTGAATTACGGACATTCAATCATCACCACGCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai Hu et al.
Oncotarget, 8(20), 33265-33275 (2017-04-13)
Bile acids (BAs) was critical in the initiation and progression of various tumors. However, their prognostic significance in pancreatic cancer was still illusive. In the present study, the expression and biological significance of FXR, a major receptor of BAs, in
Wenxuan Xu et al.
IUBMB life, 68(5), 376-387 (2016-03-31)
Hepatic stellate cells (HSCs) are universally acknowledged to play a stimulative role in the pathogenesis of hepatic fibrosis and portal hypertension. HSCs when activated in response to liver injury are characterized with many changes, with HSC contraction being the most
Wenxuan Xu et al.
Toxicology and applied pharmacology, 315, 23-34 (2016-12-13)
Alcoholic liver disease (ALD) is a common etiology of liver diseases, characterized by hepatic steatosis. We previously identified farnesoid X receptor (FXR) as a potential therapeutic target for ALD. Dihydroartemisinin (DHA) has been recently identified to possess potent pharmacological activities
Li-Min Liang et al.
Acta pharmacologica Sinica, 41(11), 1465-1475 (2020-04-29)
Peptide transporter 1 (PepT1), highly expressed on the apical membrane of enterocytes, is involved in energy balance and mediates intestinal absorption of peptidomimetic drugs. In this study, we investigated whether and how diabetes affected the function and expression of intestinal
Renchao Dong et al.
European journal of pharmacology, 857, 172461-172461 (2019-06-21)
Estrogen-induced cholestasis is a common etiology of hepatic diseases in women with contraceptives administration, pregnancy or hormone replacement therapy. Farnesoid X receptor (FXR) is a member of nuclear receptor super family of ligand-activated transcription factors that is highly expressed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service